|
The time and effort expended on optimizing the growth conditions of recombinant organisms is amply rewarded with convenient fermentation and high yield of recombinant protein. After the expression system has been determined, and the preferred growth medium selected, it is critical to craft a set of conditions that results in low stress and high productivity. It's not surprising: combine agitation with surfactant and you get ... foam. In any suspension culture whether your protein expression host is bacterial, yeast, insect, or mammalian the growth medium needs to be agitated to aerate and suspend the cells. And many culture media contain proteins and other components with surfactant properties. Whenever these two conditions are met, foam is an almost inescapable complication. Why should you care about foam? There are several good reasons:
What to do? | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| Antifoams |
Admittedly, this isn't rocket science. Truth be told, it's rather a low-tech answer to your high-tech problem. But it is an answer; it works; and it has the virtue of simplicity. What's an antifoam?Antifoams have been used in foodstuffs, pharmaceuticals, and manufacturing processes for many years. They are silicone-based polymers, non-silicone organics, or mixtures of both. They work at very low concentrations to suppress foaming in aqueous media, typically in the range of 0.005 0.02%. Here are a few things you should know when using antifoams in cell culture systems:
It's as simple as that. Sigma's antifoams (see Table 1) all have been tested with E. coli (ATCC #25922) and B. subtilis (ATCC #6051) in Terrific Broth. In all cases, the final OD600 in cultures with the antifoam is at least 80% of the value without it. We're in the process of extending testing to other expression systems and other media. If you check back here, on our Web site, you'll be among the first to know. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Table 1: Which Antifoam is Right for
Your Needs?
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| Keeping it Clean: A New Approach to Preparing Microbial Media with EZMix |
While we're on the subject of components for efficient growth of recombinant organisms, let's take a look at media. For the best results, you must provide a rich, complete growth medium.This usually involves a standard-recipe broth with or without specific supplementation to meet the nutritional requirements of the expression host. Though it is one of the most fundamental techniques in microbiology, media prep can go awry. Cleanliness is key. Contamination, wasted time and materials, and confused results are all but inevitable without it. Yet most microbial media components are dusty, sticky powders hard to pour, hard to weigh, hard to dissolve making it that much more challenging to meet the cleanliness commitment. We'd like to propose a solution. At Sigma, we developed a proprietary process that converts standard microbial media components into EZMix free-flowing, non-dusting, dense, wettable powders. Each EZMix powder has the same growth characteristics as the untreated powder, with clear advantages in handling and dissolution. These EZMix media and media components are the first in a growing family. Though the EZMix process is a benign one, we take nothing for granted. Each new formulation is tested before release so you can be assured that it will perform as expected. If your favored medium is not yet on this list or if you have some comments about your experiences with EZMix powders, drop us an e-mail. We want to hear from you. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| Table 2: Non-dusting EZMix Products |
| ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| Expression
Solutions: Questions and Answers |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Rather than produce a
sterile stock solution at the outset, can I add the antifoam to the medium and
autoclave it all at once? |
Yes you can. This can be a
convenient method when preparing small aliquots of sterile medium. However, you
might also want to keep a sterile stock solution available in case you need to
add more antifoam while the culture is growing. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Are antifoams the same
as defoamers, or are we talking about two different things here? |
Antifoams and defoamers are
very similar chemically. The main differences lie in when and how they are used.
A defoamer is sprayed or dispersed over an already foamy surface, and should
work to eliminate foam quickly, but won't necessarily prevent refoaming after it
dissolves. An antifoam is introduced into the foam-prone medium, often before
any foam appears, and it should prevent foam from forming as long as it is
present in the proper concentration. The concentration at which it's used,
though, won't always defoam an already foamy solution at least not very
quickly.
| ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
My fermentation
protocols are already well optimized. What kind of risks am I running if I
switch to your EZMix media formulations? |
Frankly, we don't see a down
side. We can assure you that the EZMix process provides the same growth
characteristics as the untreated powder because we test each new formulation
with both E. coli and B. subtilis. We don't release any
formulation that fails to deliver at least equal growth rates and
maximum ODs when compared to the untreated powder. Sometimes, we actually see
slightly better growth with EZMix! | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
I have long experience
with my old favorites like Terrific Broth and LB broth. Are you sure EZMix
versions can offer the same performance that I'm accustomed to? |
Actually, not the same
better! In developing the EZMix products, we set tighter product
specifications. The EZMix formulations can be relied upon to perform at the same
high levels, every time, lot after lot. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EZMix is a trademark of Sigma-Aldrich Co.