Extranet 
Welcome to SAFC  Biosciences
Serum Free Media for Insect Cell Culture
EX-CELL™ Insect Expression Products
 On this Page
Grace's Insect Medium
Hink's TNM-FH
IPL-41
TC-100
Table bottom
Grace's Insect Medium
Catalog # Name
56902C Grace's Insect Medium
Original Formula, with 4.1 mM L-glutamine, without calcium chloride, without sodium bicarbonate
Dry Powder Medium (DPM)
  > Grace's Insect Medium
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 1 L, 5 L, 10 L, 50 L, 100 L
  Storage Conditions: 2 to 8 °C
51902C Grace's Insect Medium
Vaughn's Modified, with 4.1 mM L-glutamine
Liquid Medium (LM)
  > Grace's Insect Medium
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 500 mL, 1000 mL
  Storage Conditions: 2 to 8 °C
56910C Grace's Insect Medium
Vaughn's Modified, with 4.1 mM L-glutamine, without sodium bicarbonate
Dry Powder Medium (DPM)
  > Grace's Insect Medium
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 1 L, 5 L, 10 L, 50 L, 100 L
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top
Hink's TNM-FH
Catalog # Name
51942C Hink's TNM-FH Insect Medium
supplemented Grace's, with 4.1 mM L-glutamine, with 3.33 g/L lactalbumin hydrolysate (LAH), with 3.33 g/L ultrafiltered yeastolate
Liquid Medium (LM)
  > Hink's TNM-FH Insect Medium
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 500 mL, 1000 mL
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top
IPL-41
Catalog # Name
56923C IPL-41 Insect Medium Modified
with 6.8 mM L-glutamine, without calcium chloride, without sodium bicarbonate
Dry Powder Medium (DPM)
  > IPL-41 Insect Medium Modified
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 1 L, 5 L, 10 L, 50 L, 100 L
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top
TC-100
Catalog # Name
56941C TC-100 Medium
with 4.1 mM L-glutamine, without sodium bicarbonate
Dry Powder Medium (DPM)
  > TC-100 Medium
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 1 L, 5 L, 10 L, 50 L, 100 L
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top