Extranet 
Welcome to SAFC  Biosciences
Disposables
Disposable Bioprocessing Systems
BIOEAZE® single-use systems offer a wide range of catalog products as well as the ability to make custom bags, bioreactor kits, sample, rigging and tubing sets as needed to meet client’s requirements. Catalog product is off-the-shelf and ready to go, or we can design and supply custom systems with the fastest turnaround time in the market.
Product Details:
   cGMP manufactured, ISO Class 13485 in a class 10,000 certified clean room
   Class VI bag films, components for incorporation into designs as selected
   Bags from 50 mL to 2,000 L, Polyethylene (PE) and Ethyl Vinyl Acetate
 (EVA) films
   Bag assemblies, rigging sets, filter sets, components, tank liners, tubing,
 storage and transport
   Custom systems designed to your specifications
   Technical knowledge, expertise and support when you need it
Benefits:
There are real benefits associated with BIOEAZE® single-use systems.
   Safety – management of contamination risk
   Speed – reduction and management of lead times
   Scaleable – bags and films from 50 mL to 2,000 L
   Cost management – supports facilities’ growth strategy, as single-use systems
 replace stainless steel and glass
   Time efficiency – systems are provided assembled and sterile
   Versatility and innovation – BIOEAZE® is completely customized to fit
  your process and enhance your operations
   Success – BIOEAZE® is currently used in the manufacture of approved  pharmaceutical products
   Minimizes cleaning validation
   Plays to rapid scale-up and scale-back
   Saves space
System Design
BIOEAZE® bioprocessing systems can be designed for your specific facility and liquid handling needs. By contacting your Business Development Manager or Customer Service Representative, you have direct access to our BioProcess Engineers (BPE). Once a request is made, our BPE group can either work with a drawing you have provided or design a system around your exact needs. To ensure that your specifications are met, all final drawings require your final sign-off and approval before manufacture.
Design Process
   Consultation with BPE
   SAFC Biosciences drawing developed
   Customer drawing reviewed, revised and signoff
   Specification developed, reviewed and signoff
   SAFC Biosciences drawing developed
   Package complete and ready to be ordered
Testing and Technical Support
SAFC Biosciences provides a wide range of testing and validations, which will support you throughout the entire development and manufacturing process.
Our single-use bioprocessing systems are tested to stringent specifications to ensure that our customers are supplied with the highest quality products possible. 
For details on testing and technical information please click on the Related Files link. Documents in this section include, but are not limited to:
   Film Information
   Testing Biological – USP Class VI, Endotoxin Physical – Structure,
Permeability Functional – Irradiation Validation, Shipping integrity
In-Process – Raw Materials and Manufacturing Inspections
   Technical Bulletins
   Definitions
A detailed validation binder is available upon request.
Related Disposable Links
Related Documents
» BIOEAZE® bags-Ethyl Vinyl Acetate(EVA) film
- T081 (82 KB pdf)
» Chemical Compatibility for Bag Films
- B008 (55 KB pdf)
» Long-term Stability of Liquid Cell Culture Media in EVA and PE Bags
- T069 (90 KB pdf)