Extranet 
Welcome to SAFC  Biosciences
Disposables Products
Disposables Products
Product Overview
Disposable Bioprocessing Systems
BIOEAZE® single-use systems offer a wide range of catalog products as well as the ability to make custom bags, bioreactor kits, sample, rigging and tubing sets as needed to meet client’s requirements. Catalog product is off-the-shelf and ready to go, or we can design and supply custom systems with the fastest turnaround time in the market.
Product Details:
   cGMP manufactured, ISO Class 13485 in a class 10,000 certified clean room
   Class VI bag films, components for incorporation into designs as selected
   Bags from 50 mL to 2,000 L, Polyethylene (PE) and Ethyl Vinyl Acetate
 (EVA) films
   Bag assemblies, rigging sets, filter sets, components, tank liners, tubing,
 storage and transport
   Custom systems designed to your specifications
   Technical knowledge, expertise and support when you need it
Benefits:
There are real benefits associated with BIOEAZE® single-use systems.
   Safety – management of contamination risk
   Speed – reduction and management of lead times
   Scaleable – bags and films from 50 mL to 2,000 L
   Cost management – supports facilities’ growth strategy, as single-use systems
 replace stainless steel and glass
   Time efficiency – systems are provided assembled and sterile
   Versatility and innovation – BIOEAZE® is completely customized to fit
 your process and enhance your operations
   Success – BIOEAZE® is currently used in the manufacture of approved
 pharmaceutical products
   Minimizes cleaning validation
   Plays to rapid scale-up and scale-back
   Saves space
BIOEAZE® Disposables Products