|
BIOEAZE® Ethyl Vinyl Acetate (EVA) bags are constructed from a single-web, medical grade laminate of four different films, with a fluid contact surface of EVA, with two layers of low-density polyethylene (LDPE) enveloping a layer of ethyl vinyl alcohol (EVOH).
BIOEAZE® Polyethylene (PE) bags are constructed with two layers (a dual-web) of translucent film, where the fluid contact surfact (the innermost layer) is a single layer of Dow ATTANE ultra low-density polyethylene (ULDPE). ULDPE offers low termperature flexibility, is non-leachable and inert, making it ideal for containing liquids. The outer layer is a laminate of nylon, polyvinylidene chloride (PVDC) and linear low density polyethylene (LLDPE). These resins provide a barrier to gas transmission and are puncture resistant.
| Bag Products |
| Product Number |
Description |
| BIOEAZE® Ethyl Vinyl Acetate (EVA) Bags |
| 1329B |
Designed for use with Terumo Aseptic Connecting Device (ACD) Applications, Double Polybagged, 20 L
|
| 1330B |
Designed for use with Quick Connect Applications, Double Polybagged, 20 L |
| 1335B |
Designed for use with Terumo Aseptic Connecting Device (ACD) Applications, Double Polybagged, 1 L |
| 1446B |
Individually Polybagged - sterile; γ-irradiated, 350 mL |
| 1521B |
Designed for use with Terumo Aseptic Connecting Device (ADC) Applications, Double Polybagged, 1 L
|
| 1523B |
Designed for use with Quick Connect Applications, Double Polybagged, 10 L |
| 1524B |
Designed for use with Terumo Aseptic Connecting Device (ACD) Applications, Double Polybagged, 10 L |
| 1564B |
Designed for use with Terumo Aseptic Connecting Device (ACD) Applications, Individually Polybagged, 350 mL
|
| 1933B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect or Luer Lock Applications, Double Polybagged, 2 L |
| 1934B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect or Luer Lock Applications, Double Polybagged, 5 L |
| 1935B |
Designed for use with Quick Connect Applications, Double Polybagged, 350 mL
|
| 1936B |
Designed for use with Quick Connect Applications, Individually Polybagged, 1 L
|
| 1937B |
Designed for use with Quick Connect Applications, Double Polybagged, 2 L
|
| 1938B |
Designed for use with Quick Connect Applications, Double Polybagged, 5 L
|
| 2338B |
Designed for use with Terumo Aseptic Connecting Device (ADC) Applications, Individually Polybagged, 350 mL
|
| 3392B |
Designed for use with the ReadyMate Disposable Aseptic Connector (DAC) or Terumo Aseptic Connecting Device (ACD), 1 L |
| 3765B |
Designed for use with Terumo® Aseptic Connecting Device (ACD) Applications, 100 mL |
| BIOEAZE® Polyethylene (PE) Bags |
| 1331B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect Applications, Double Polybagged, 50 L |
| 1332B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect Applications, Double Polybagged, 100 L |
| 1333B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect Applications, Double Polybagged, 200 L |
| 1824B |
Designed for use with Quick Connect Applications, Double Polybagged, 1000 L |
| 1946B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect Applications, Double Polybagged, 50 L |
| 1947B |
Designed for use with Terumo Aseptic Connecting Device (ACD) and Quick Connect Applications, Double Polybagged, 100 L |
| 2339B |
Designed for use with Quick Connect Applications, Double Polybagged, 500 L |
| 3398B |
Designed for use with Quick Connect Applications, Double Polybagged, 5 L |
| 3399B |
Designed for use with Quick Connect Applications, Double Polybagged, 10 L |
BIOEAZE is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
C-Flex is a registered trademark of Concept Polymer Technologies Inc.
|