Extranet 
Welcome to SAFC  Biosciences
Disposable Tank Liner Products
BIOEAZE® Tank Liners
  Tank Liner Products
 Product Number  Product Description
 2220L  Length 12.5" x Width 11.5" x Height 44.0"  
 pkg of 20 - 50L
 Tank Liner, "3D" Style
 2221L  Length 15.7" x Width 14.7" x Height 33.0"  
 pkg of 20 - 100L
 Tank Liner, "3D" Style
 2222L  Length 19.0" x Width 18.0" x Height 38.0"  
 pkg of 20 - 200L
 Tank Liner, "3D" Style
 1569L  21.0" diameter x 39" height 
 pkg of 20 - 50L
 Tank Liner, "Pillow" Style
 1570L  29.0" diameter x 48" height  
 pkg of 20 - 130L
 Tank Liner, "Pillow" Style
 1571L  36.0" diameter x 51" height 
 pkg of 20 - 200L
 Tank Liner, "Pillow" Style
BIOEAZE is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
Have a question or just curious?
BIOEAZE® Disposables Products