Extranet
SAFC Global
SAFC Buchs - Kanton St. Gallen, Switzerland
SAFC Pharma - SAFC Supply Solutions
SAFC Buchs has a long history of producing high-quality intermediates and bulk actives.The plant specializes in complex, multi-step synthesis to 1,600 liters and low temperature reactions (-60°C) up to 400 liters. A broad range of production technologies and multi-purpose equipment offers batch sizes from grams to tons. This facility operates under cGMP, certified by the Swiss authorities.
SAFC Buchs: cGMP manufacturing site for organic fine chemicals.
What our technologies can provide for you
Custom synthesis and process development
Production of high-quality intermediates and bulk actives
Technology strengths are aimed toward low-volume, highly complicated chemistries
Reactor capacity from 5 to 400 gallon (20 L - 1,600 L) glass-lined and Hastelloy
Operating temperature range from -60°C up to +180°C
Safety of all processes is evaluated using state-of-the-art DSCRC 1 and ARC calorimeter equipment
Contact SAFC Buchs
SAFC Buchs Facility in Kanton St. Gallen, Switzerland
  Quality Manager
Industriestrasse 25
9470 Buchs
Switzerland
Swiss Free Call: 0800 80 00 80
Tel: +41 (0) 81 755 27 32
Fax: +41 (0) 81 755 27 70
Schedule a Site Audit
Request Quality Information
View Site Overview
View Expertise Chart
View Virtual Tours of facility
Certificates and Documents
cGMP certificate from Swiss Authorities
(English PDF)
cGMP certificate from Swiss Authorities
(German PDF)
SQS ISO 9001:2000
59k jpg
ISO 9001:2000
60k jpg