Extranet 
Welcome to SAFC HiTech
Analytical Services
SAFC Hitech - Compound Semiconductor
To support production and R&D operations, our state-of-the-art tools provide analytical testing to the highest sensitivities for a wide range of contaminants. We offer an extensive array of techniques to fully characterize compounds and measure impurities to the lowest of levels.
For example, primary techniques employed for analysis of products to the semiconductor industry include FT-NMR, ICP-OES/MS, HR-ICP-MS and GC, but many more are employed to provide characterization or additional data on each product, including TGA, vapor pressure measurement and particle counting, depending upon customer requirements. SAFC Hitech has developed unique handling and sample preparation procedures that, coupled with cleanroom environments, ensure best performance from analytical tools utilized. Such capabilities are critical to ensuring background levels of contamination are minimized to achieve the low detection limits required.
Analytical instruments are operated by a team of skilled and highly trained analytical chemists. Instruments are validated using tools such as Gage R&R to monitor performance.  
Monitoring of advances in analytical technology and review of methodology are continuously employed the analytical capability can be improved to meet future expectations and challenges for product or specification development and improvement.
Our range of analytical services includes:
top curve blue
Identity Testing
Purity Testing
Identity Testing
Appearance
All compounds that pass through our quality department are automatically assigned this test code. This is often the first test that customers perform without realizing it. Although simple in nature, it can reveal many subtle flaws in the compound.
Spectroscopy
IR
· We employ both NIR and ATR. Both methods have different strengths and allow fundamental analysis of compounds to identify the compound or give clues to some of the impurities.
Multinuclear NMR (NMR)
· SAFC has a wide level of experience on several different isotopes- including but not limited to H, 13C, P, S, B, and F.
UV/Vis
Fluorescence
Mass Spectra
· Some sources include Chemical Ionization (CI), Electron Impact (EI), Electrospray Ionization (ESI), Atmospheric Pressure Chemical Ionization (APCI), and Matrix Assisted Desorption Ionization-Time of Flight (MALDI TOF)
RAMAN
Elemental Analysis
Carbon/Hydrogen/Nitrogen/Sulfur (CHNS)
Purity Testing
Chromatography
SAFC has a very wide range of analytical capibility in this area.
Gas Chromatography (GC)
· Thermal Conductivity Detector (TCD)
· Electron Capture Detector (ECD)
· Flame Ionization Detector (FID)
· Mass Spectra (MS)
Liquid Chromatography (LC)
· Ultra Violet Detector (UV)
· Mass Spectra (MS)
· Evaporative Light Scattering Detector (ELSD)
· Electrochemical Detector (ECD)
· Fluorescence Detector (FLD)
· Refractive Index (RI)
Ion Chromatography
Thin Layer Chromatography
Differential Scanning Calorimetry (DSC)
Thermogravimetric Analysis (TGA)
Spectroscopy
Flame Atomic Absorbtion (AA)
Graphite Furnace Atomic Absorption (GFAA)
Inductively Coupled Plasma Optical Emission Spectra (ICP-OES)
Inductively Coupled Plasma Mass Spectroscopy (ICP-MS)
Magnetic Sector Field Inductively Coupled Plasma Mass Spectroscopy (HR-ICP-MS)
Water Content Analysis
Karl Fischer Coulometric Titration
Karl Fischer Volumetric Titration
Elemental Analysis
Carbon/Hydrogen/Nitrogen/Sulfur (CHNS)
Density
Polarography
Vapor Pressure Measurement
Particle counting Measurement