Extranet 
Welcome to SAFC HiTech
 Environmental Health & Safety
SAFC Hitech® — Overview
"SAFC Hitech is committed to protecting the environment and the health and safety of all our employees, their families, our customers and the public, and is dedicated to being a leading company in health, safety and environmental issues by continuously improving our performance to benefit employees, customers, shareholders and the community alike."
— A.B. Leese President, SAFC Hitech
As a manufacturer of hazardous chemicals, all aspects of health and safety are of the utmost importance to enable continuity of supply to our valued customers. Our safety philosophy is not merely a statement but a role model for all of our employees to follow. To achieve this goal, a number of techniques are integrated into our manufacturing capabilities and processes.
1. Hazard and Operability Studies (HAZOP)
HAZOPs identify possible causes of deviation, what the consequences might be and whether the correct safety feature can be installed or is present. The process is normally introduced at the plant design stage by a multi-disciplined team of professionals.
2. Fault Tree Analysis (FTA)
FTA is primarily designed for product failure and trouble shooting and is characterized by each area of analysis creating its own chain or flow resulting in a fault being identified.
3. Risk Assessment
All tasks follow a risk assessment procedure with the sole purpose of identifying hazards and ensuring control measures are in place to reduce the hazards to an acceptable level.
4. Management of Change (MOC) Procedure
All installations and modifications to the plant and processes have to pass a rigorous procedure of authorization through our MOC process, ultimately resulting in acceptance of legal standards and practices prior to implementation and operation.
5. "What If"
All relevant procedures within our ISO 9002 accredited system carry a "what if" section which subsequently advises operatives as to the action required to eliminate any unexpected problems.
6. Control
All Health & Safety aspects of the company business are audited to ensure compliance with legislative requirements, and to ensure employees are conforming to set procedures.
7. General
As a principle, we actively encourage participation in national working groups engaged in setting occupational sector standards endorsed by national policing authorities. Currently, SAFC Hitech employees are involved in many committees including:
i) Chemical Industry Association
ii) Health and Safety Executive Microelectronics Semiconductor Manufacturing Group
iii) Local Environmental Planning and Control
iv) Responsible Care Group
Our aim is to manufacture and supply products which all parties are confident of using, handling and storing with peace of mind and the knowledge that, within every aspect of the supply chain, the safety of everyone is of prime importance and endorsed by the high standards we set.
General Safety Advice
  • Only experienced and correctly trained persons should handle our products
  • Observe all regulations and local requirements regarding handling and storage
  • Do not deface labels provided with the product
  • Know and understand the properties and hazards associated with the product
  • Before using any product ensure you are familiar with emergency situations that may arise
  • Always use the correct protective clothing as identified on the Material Safety Data Sheet
  • If you have any doubt as to the correct handling procedures, contact an SAFC Hitech representative
Key to Risk and Safety Phrase Codes used in Health and Safety Guide
Risk Phrases (R)
10: Flammable
11: Highly Flammable
12: Extremely Flammable
14: Reacts violently with water
17: Spontaneously flammable in air
23: Toxic by inhalation
24: Toxic in contact with skin
25: Toxic if swallowed
26: Very toxic by inhalation
34: Causes burns
35: Causes severe burns
36: Irritating to the eyes
37: Irritating to the respiratory system
38: Irritating to the skin
40: Possible risk of irreversible effects
45: May cause cancer
48: Danger of serious damage to health by prolonged exposure
50: Very toxic to aquatic organisms
52: Harmful to aquatic organisms
53: May cause long term adverse effects in the aquatic environment
Safety Phrases (S)
6: Keep under inert atmosphere
8: Keep container dry
9: Keep container in a well ventilated place
16: Keep away from sources of ignition - No smoking
20: When using do not eat or drink
23: Do not breathe fumes
24: Avoid contact with the skin
25: Avoid contact with the eyes
26: In case of contact with eyes, rinse immediately with plenty of water and seek medical advice
28: After contact with skin, wash immediately with plenty of water
36: Wear suitable protective clothing
37: Wear suitable gloves
38: In case of insufficient ventilation, wear suitable respiratory equipment
39: Wear eye/face protection
43: In case of fire use dry powder or lime - Never use water
45: In case of accident or if you feel unwell seek medical advice immediately. (Show label where possible)
53: Avoid exposure - obtain special instruction before use
59: Refer to manufacturer for information on recycling/recovery
60: This material and/or container must be disposed of as hazardous waste
61: Avoid release to the environment. Refer to special instructions/safety data sheet