Extranet 
Welcome to SAFC Pharma
Development and Manufacture of Therapeutic Proteins through Fermentation Technology
SAFC Pharma®
Therapeutic Proteins - Development and Manufacture through Fermentation Technology ImageTherapeutic proteins are a fast growing segment of the pharmaceutical industry. These include native proteins, recombinant and fusion proteins, peptides and antibody fragments, many of which are manufactured through microbial fermentation. Building on 30 years of experience, SAFC Pharma offers contract development and manufacturing of fermentation-derived therapeutic proteins from pre-clinical to commercialization.
Microbial Systems
SAFC Pharma employs microorganisms up to Risk Group 2 and operates at Bio-safety level 2 large scale. Project specific master and working cell banks are safely deposited in temperature-controlled bio-storage. Strains, handled under strict aseptic procedures, are identified and characterized for homogeneity (absence of foreign growth), viability and productivity. Native or recombinant proteins may be recovered from the fermentation broth (secreted) or the biomass (e.g. intracellular, periplasmic, soluble, or in inclusion bodies). Yields are optimized through both upstream and downstream process development. SAFC Pharma has broad experience in microbial fermentation, including but is not limited to:
Red Square Bullet   Bacteria (e.g. Escherichia coli, Staphylococcus aureus, Clostridium species)
Red Square Bullet   Streptomycetes (e.g. Streptomyces species, Actinomyces species)
Red Square Bullet   Filamentous Fungi (e.g. Nigrospora species, Aspergillus species)
Red Square Bullet   Yeast (e.g. Saccharomyces cerevisiae, Pichia pastoris)
Manufacturing Capabilities
Red Square Bullet   The state-of-the art GMP fermentation facility in Jerusalem, Israel features:
   Fully automated equipment
   Cleaning and sterilization/sanitization in place (CIP, SIP) systems
   Sanitary grade piping equipped with filters
   Uniform fermentor geometry and pilot/ production purification system consistency to ensure robust and scalable
processes
Red Square Bullet   Class 100,000 (ISO 8) Manufacturing suite:
   Fermentors - 100 L / 1,000 L / 4,000 L
   Continuous disc stack centrifuge
   Automatic piston discharge high-speed centrifuge
   High-pressure homogenizer
   Temperature-controlled, stirred processing tanks up to 4,000 L
Red Square Bullet   Class 10,000 (ISO 7) segregated purification suites:
   GE Uniflux™ tangential flow ultra/diafiltration, Unicorn™ software
   Millipore® Liquid BioChromatography production and pilot-scale systems
   BPG QuickScale high-performance columns
Red Square Bullet   Lyofast 5 pharmaceutical freeze dryer (Lyophilizer)
Quality Management
Dedicated Quality Assurance (QA) and regulatory affairs personnel are in place to handle:
Red Square Bullet   Document review and approval (e.g. MMF, SOPs, batch records, product release)
Red Square Bullet   Equipment qualification including 21CFR part 11 compliance
Red Square Bullet   Validation of manufacturing processes and cleaning procedures
Red Square Bullet   Investigation of non-conformances
Red Square Bullet   Change control implementation
Red Square Bullet   Corrective And Preventive Action (CAPA)
Red Square Bullet   Personnel training and qualification program
Red Square Bullet   Drug Master File (CTD-DMF) filing with the regulatory agencies (e.g. FDA, EMEA)
Analytical Services
Therapeutic proteins analytical tests are compliant with Pharmacopoeia (EP/USP) methods and ICH guidelines. Managed via a validated Informatics System for Quality Control (QC) analytical data (SAP-QM), our services include:
Red Square Bullet   Analytical methods development and validation
Red Square Bullet   Testing of raw materials, in-process control, and final product release
Red Square Bullet   Stability testing
Red Square Bullet   Cleaning efficiency testing
Red Square Bullet   Selected protein analysis tests:
Test Methods
Protein concentration Biuret, Lowry, UV (A280), W/C, Bradford
Molecular mass ESI-MS
Protein identity Western blot, LC-MS (Peptide mapping), Amino acid analysis
Protein purity RP-HPLC, TLC, SDS-PAGE
Immunoassay ELISA, Radioimmunoassay, Immunofluorescence
Potency/Activity Bio assays (Enzymatic, Affinity, Attachment, Lytic)
Impurities Heavy metals, Water (KF)
Residuals qPCR (DNA), LC-MS/MS (Kanamycin), HPLC (IPTG)
Endotoxin LAL
Microbial load Bio-burden, Sterility
>> Download the print version of this page Fermentation Insert (1.08 Mb PDF)
>> View the Israel Facility
Request Information