|
Custom Formulation and Fill / Finish
 SAFC Pharma® provides fill/finish services for biologic drug products as well as small-molecule APIs up to 10,000 vials per lot. Our highly trained teams have a service record of more than 500,000 vials and syringes filled for over 300 lots without a single sterility failure.
| Semi-Automated and Fully-Automated Fills |
| |
Capacity: up to 10,000 vials/lot |
| |
0.15-23 mL fill volumes |
| |
0.3-30 mL vial size |
| |
Typical turnaround of 10 weeks (plus testing) - expansion possibilities based on customer demand |
| |
Glass vials, cryovials, syringes and bags available |
| |
Final product, diluents, placebo |
| |
Labeling options for fully-blinded studies |
| |
Automated labeling equipment |
| Key Features of the Filling Operations |
| |
Strict accordance to cGMP standards, with robust quality systems that ensure every fill is performed according to specification |
| |
Highly-experienced personnel |
| |
Optimized documentation systems for efficient project turn-around |
| |
On-site storage |
| |
IATA certified shipping |
| Facility suited for FDA and EU-regulated clinical trials and features |
| |
BSL2 (BioSafety Level 2) enhanced containment |
| |
Class 1,000 (ISO 6) suites and Class 100 (ISO 5) Biosafety cabinets |
| |
100% single-pass HEPA filtered air |
| Quality Management |
| |
Dedicated quality assurance (QA) teams |
| |
Highly experienced specialists |
| |
Clinical product successfully filled for trials in US, EU and Asia |
| |
On-site storage |
| |
Rigorous compliance program |