Extranet 
Welcome to SAFC Pharma
SAFC Pharma® – St. Louis Facility
Biologic APIs and Conjugation
SAFC St. Louis – Missouri (USA)
SAFC Pharma's St. Louis, Missouri (USA) campus is a key center of manufacturing excellence with 25 years experience in cGMP biologics.
   Plant based therapeutic protein extraction and purification
   Animal-based therapeutic protein extraction and purification
   Excipients and adjuvants
   Bio-organic APIs synthesis and purification
   Bioconjugation
   High potent bioconjugation
SAFC St. Louis – Missouri (USA)
>> For more information view the St. Louis Facility Insert
Facility Overview:
  Completely segregated manufacturing operations
  Large-scale capacity to extract biomass at up to 6,500 kg/week up to
  Purification in 1.5 m of diameter chromatography columns
  Extensive analytical capabilities and dedicated compliance
Animal-sourced APIs Manufacturing
Purpose-built, completely segregated facilities have been specifically designed to support animal protein extraction and purification at commercial scale, to meet growing customer demand for animal-sourced therapeutic proteins.
Manufacturing for sample projects includes:
  Thrombin (wound care)
  Hyalruonidase (eye care)
  Porcine/bovine sourced Trypsin
  Wide variety of excipients (such as Keyhole Limpet Hemocyanin)
St. Louis Pipeline
  Plant-sourced APIs
Dedicated upstream plant biomass extraction and clarification facility equipped to process up to 6,500 kg of plant biomass/week.
Facility features:
  Separate cGMP plant extraction and downstream purification
  Purification ISO 7 and ISO 8 classified suites
  Unidirectional process flows
  Equipment wash room
  Fixed, stainless steel jacketed biopharm grade tanks to 3,200 L
  Disposable technology availability
  Delta V control system, central Clean In Place (CIP) skid chromatography and TFF skids
Bio-organic APIs Synthesis and Purification
SAFC Pharma offers a wide range of bio-organic cGMP APIs, adjuvants, excipients and intermediates serving both the pharmaceutical and biopharmaceutical industries.
Manufactured from both natural and synthetic sources, our offer includes:
   Lipids
   Detergents
   Buffers
   Polyamino Acids
   Carbohydrates
   Custom solvent sanitization blends
Our fully validated facility offers reactor and crystallization capacity up to 3,000 gallons and vacuum filtration capability of up th 0.6 square meters for up to 100K kg of finished product batches within a temperature range of –20 °C to 120 °C.
Potent and Non-Potent Conjugation
To support exciting new cancer drugs, SAFC Pharma is one of the few companies to offer the crucial combination of small-molecule chemistry, biologics capabilities and high-containment engineering to bring HPAPI conjugates to commercialization.
Non-potent conjugates, primarily used for targeted drug delivery applications, are also available.
The new cGMP HPAPI conjugation facility features:
   800 sq. ft. floor space
   ISO 8 (Class 100,000) room classification
   Walker™ dual-sided six glove isolator
   10-100 L reactor size
   Batch size capability over 100 g
Go to SAFC Pharma's Webpage Virtual Tours Available
 cGM Protein Extraction & Purification
 cGMP Manufacturing
 cGMP Bio-organic Manufacturing
 See all virtual tours