|
4-Chloro-N-Methylpiperidine is an advanced intermediate used in the manufacture of a large number of Active Pharmaceutical Ingredients. It is avaiable as the free base or the hydrochloride salt.
4-Chloro-N-Methylpiperidine has been manufactured at the SAFC Arklow facility since 1989.
• Flexible packing 5-200 kgs in mild steel or plastic drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
4-Chloro-N-Methylpiperidine |
| Chemical Formula |
C6H12ClN |
| M.W. |
133.62 |
| Chemical Name |
4-Chloro-1-methylpiperidine |
| CAS Number |
5570-77-4 |
| EINECS Number |
226-942-4 |
|