|
8-Chlorotheophylline is a xanthine derivative with stimulant properties. It is combined with pharmaceutical drugs to form stable salts, such as dimenhydrinate.
8-Chlorotheophylline has been manufactured at the SAFC Arklow facility since 1986.
• Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
8-Chlorotheophylline |
| Chemical Formula |
C7H7ClN4O2 |
| M.W. |
214.61 |
| Chemical Name |
8-Chloro-1,3-dimethyl-2,6(1H,3H)-purinedione |
| CAS Number |
85-18-7 |
| EINECS Number |
201-590-4 |
|