|
Acepromazine or Acetylpromazine (more commonly known as ACP, Ace, or by the trade names Atravet or Acezine 2, number depending on mg/ml dose) is a phenothiazine derivative antipsychotic drug. It was first used in humans in the 1950s, but is now little used in humans (the closely related analogue, chlorpromazine, is still used as an antipsychotic in humans), but frequently in animals as a sedative and antiemetic.
Acepromazine Maleate has been manufactured at the SAFC Arklow facility since 1989. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Acepromazine Maleate |
| Chemical Formula |
C19H22N2OS · C4H4O4 |
| M.W. |
442.50 |
| Chemical Name |
10-(3-Dimethylaminopropyl)phenothiazine-2-yl Methyl Hydrogen Maleate |
| CAS Number |
3598-37-6 |
| EINECS Number |
222-748-9 |
|