|
Amitriptyline (or Amitryptyline) hydrochloride (sold as Elavil, Tryptanol, Endep, Elatrol, Tryptizol, Trepiline, Laroxyl, Saroten, Triptyl, Redomex) is a tricyclic antidepressant drug. It is a white, odorless, crystalline compound which is freely soluble in water; it is usually dispensed in tablet form.
Amitriptyline Hydrochloride has been manufactured at the SAFC Arklow facility since 1988. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Amitriptyline Hydrochloride |
| Chemical Formula |
C20H23N · HCl |
| M.W. |
313.87 |
| Chemical Name |
3-(10,11-Dihydro-5H-Dibenzo[a,d]cyclopenten-5-ylidine)propyldimethylamine Hydrochloride |
| CAS Number |
549-18-8 |
| EINECS Number |
208-964-6 |
|