|
Brompheniramine (also known as Bromfed, Dimetapp, Bromfenex, and Dimetane), commonly marketed as its salt brompheniramine maleate is an antihistamine drug of the propylamine (alkylamine) class.
Brompheniramine Maleate has been manufactured at the SAFC Arklow facility since 1990. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Brompheniramine Maleate |
| Chemical Formula |
C16H19BrN2 · C4H4O4 |
| M.W. |
435.31 |
| Chemical Name |
3-(4-Bromophenyl)-N,N-dimethyl-3-(2-pyridyl)propylamine Hydrogen Maleate |
| CAS Number |
980-71-2 |
| EINECS Number |
213-562-9 |
|