|
Butoconazole is an imidazole antifungal used in gynecology.
Butoconazole Nitrate has been manufactured at the SAFC Arklow facility since 2001. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Milled and Micrionised material available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Butoconazole Nitrate |
| Chemical Formula |
C19H17Cl3N2S · HNO3 |
| M.W. |
474.80 |
| Chemical Name |
(±)-1-[4-(p-chlorophenyl)-2-[(2,6-dichlorophenyl) thio]butyl] imidazole mononitrate |
| CAS Number |
64872-77-1 [mononitrate] |
| EINECS Number |
Not listed |
|