|
Clomipramine (brand-name Anafranil) is a tricyclic antidepressant. It was developed in the 1960s by the Swiss drug manufacturer Geigy (now known as Novartis) and has been in clinical use worldwide for decades.
Clomipramine Hydrochloride has been manufactured at the SAFC Arklow facility since 1988. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Clomipramine Hydrochloride |
| Chemical Formula |
C19H23ClN2 · HCl |
| M.W. |
351.32 |
| Chemical Name |
3-(3-Chloro-10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyldimethylhylamine Hydrochloride |
| CAS Number |
17321-77-6 |
| EINECS Number |
241-344-3 |
|