|
Diethylcarbamazine (DEC) is an anthelmintic drug that does not resemble other antiparasitic compounds. It is a synthetic organic compound which is highly specific for several parasites and does not contain any toxic metallic elements.
Diethylcarbamazine Citrate has been manufactured at the SAFC Arklow facility since 1986. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
8-Chlorotheophylline |
| Chemical Formula |
C10H21N3O · C6H8O7 |
| M.W. |
391.40 |
| Chemical Name |
N,N-Dimethyl-4-methylpiperazine-1-carboxamide Dihydrogen Citrate |
| CAS Number |
1642-54-2 |
| EINECS Number |
216-696-6 |
|