|
Doxylamine succinate is one of the many sedating antihistamines used by itself as a short-term sedative, in combination with other drugs as a night-time cold and allergy relief drug.
Doxylamine Succinate has been manufactured at the SAFC Arklow facility since 1986. It conforms to the USP, EP and BP specifications.
• Certificate of Suitability (COS) and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Doxylamine Succinate |
| Chemical Formula |
C17H22N2O · C4H6O4 |
| M.W. |
388.46 |
| Chemical Name |
N,N-Dimethyl-2-[α-methyl-α-2-pyridinyl)benzyloxy]ethylamine Hydrogen Succinate |
| CAS Number |
562-10-7 |
| EINECS Number |
209-228-7 |
|