|
Imipramine (sold as Antideprin, Deprenil, Deprimin, Deprinol, Depsonil, Dynaprin, Eupramin, Imipramil, Irmin, Janimine, Melipramin, Surplix, Tofranil) is an antidepressant medication, a tricyclic antidepressant of the dibenzazepine group.
Imipramine is mainly used in the treatment of major depression and enuresis. Imipramine Hydrochloride has been manufactured at the SAFC Arklow facility since 1986. It conforms to the EP and BP specifications.
• CTD available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Imipramine Hydrochloride |
| Chemical Formula |
C19H24N2 · HCl |
| M.W. |
316.87 |
| Chemical Name |
3-(10,11-Dihydro-5H-dibenz[b,f]azepin-5-yl)propyldimethylamine Hydrochloride |
| CAS Number |
113-52-0 |
| EINECS Number |
204-030-7 |
|