|
Lofepramine (trademarked Gamanil, Tymelyt, generic Lomont) is a third generation tricyclic antidepressant used in the treatment of depressive disorders. It has both antidepressant and anxiolytic properties.
Lofepramine Hydrochloride has been manufactured at the SAFC Arklow facility since 1990. It conforms to the EP and BP specifications.
• CTD available • Flexible packing 1-25 kgs in plastic drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Lofepramine Hydrochloride |
| Chemical Formula |
C26H27ClN2O · HCl |
| M.W. |
455.43 |
| Chemical Name |
5-(3-[N-(4-Chlorophenacyl)-N-methylamino]propyl)-10,11-dihydrodibenz[b,f]azepine Hydrochloride |
| CAS Number |
26786-32-3 |
| EINECS Number |
248-002-2 |
|