|
Nortriptyline is a second-generation tricyclic antidepressant marketed as the hydrochloride under the trade names Sensoval, Aventyl, Pamelor, Norpress, Allegron and Nortrilen.
Nortriptyline Hydrochloride has been manufactured at the SAFC Arklow facility since 1989. It conforms to the USP, EP and BP specifications.
• CTD and US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Nortriptyline Hydrochloride |
| Chemical Formula |
C19H21N2 · HCl |
| M.W. |
299.80 |
| Chemical Name |
3-(10,11-Dihydro-5H-Dibenzo[a,d]cyclohepten-5-ylidine)propylmethylamine Hydrochloride |
| CAS Number |
894-71-3 |
| EINECS Number |
212-973-0 |
|