|
Orphenadrine (sold under the brand names Norflex, Mephenamin, Disipal, Banflex, Flexon, Biorphen, Brocasipal, and others) is an anticholinergic drug of the ethanolamine antihistamine class with prominent CNS and peripheral actions used to treat painful muscle spasm and other symptoms and conditions as well as some aspects of Parkinson's Disease.
Orphenadrine Citrate has been manufactured at the SAFC Arklow facility since 1990. It conforms to the USP, EP and BP specifications.
• CTD available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Orphenadrine Citrate |
| Chemical Formula |
C18H23NO · C6H8O7 |
| M.W. |
461.50 |
| Chemical Name |
Dimethyl[2-(2-methylbenzhydryloxy)ethyl]amine Dihydrogen Citrate |
| CAS Number |
4682-36-4 |
| EINECS Number |
225-137-5 |
|