|
Orphenadrine (sold under the brand names Norflex, Mephenamin, Disipal, Banflex, Flexon, Biorphen, Brocasipal, and others) is an anticholinergic drug of the ethanolamine antihistamine class with prominent CNS and peripheral actions used to treat painful muscle spasm and other symptoms and conditions as well as some aspects of Parkinson's Disease.
Orphenadrine Hydrochloride has been manufactured at the SAFC Arklow facility since 1988. It conforms to the EP and BP specifications.
• CTD available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Orphenadrine Hydrochloride |
| Chemical Formula |
C18H23NO · HCl |
| M.W. |
305.90 |
| Chemical Name |
Dimethyl[2-(2-methylbenzhydryloxy)ethyl]amine Hydrochloride |
| CAS Number |
341-69-5 |
| EINECS Number |
206-435-4 |
|