|
Promethazine is a first-generation H1 receptor antagonist, antihistamine and antiemetic medication. It can also have strong sedative effects although it is rarely used specifically for this.
Promethazine Hydrochloride has been manufactured at the SAFC Arklow facility since 1988. It conforms to the USP, EP and BP specifications.
• CTD US DMF available • Flexible packing 1-25 kgs in fibreboard drums • Site is FDA and IMB inspected
Material is available ex stock for immediate shipment globally.
| Product |
Promethazine Hydrochloride |
| Chemical Formula |
C17H20N2S · HCl |
| M.W. |
320.90 |
| Chemical Name |
1,N,N-Trimethyl-2-(phenothiazin-10-yl)ethylamine Hydrochloride |
| CAS Number |
58-33-3 |
| EINECS Number |
200-375-2 |
|