|
Catalog Number: |
|
A4268 |
|
Activity: |
|
≥700 units/mg protein (E1%/280) |
|
Common Substrate(s) w Product Number(s): |
|
25590 -- maltoheptaoside 25591 -- maltotrioside N5885 -- maltoside A4640 -- Amylopectin Azure A3508 -- Amylose 10140 -- Amylose 85641 -- Starch |
|
Common Inhibitor (s) w Product Number(s): |
|
D9305 -- 1-deoxynojirimycin A3535 -- α-amylase inhibitor |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
150,000 mg |
| |
|
|
|
Catalog Number: |
|
A7011 |
|
Activity: |
|
≥300 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
459844 -- ethanol 110078 -- acetaldehyde |
|
Common Inhibitor (s) w Product Number(s): |
|
T1132 -- Tetraethyliuram disulfide |
|
Cofactor or Coenzyme: |
|
N7004 -- NAD |
|
Typical Batch Size: |
|
1,500,000 KU |
| |
|
|
|
Catalog Number: |
|
A6279 |
|
Activity: |
|
≥3 TIU/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
Trypsin inhibitor |
|
Common Inhibitor (s) w Product Number(s): |
|
|
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
200,000 ml |
| |
|
|
|
Catalog Number: |
|
A6535 |
|
Activity: |
|
ATPase ≥200 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more common: A7699 -- ATP A2754 -- ADP |
|
Common Inhibitor (s) w Product Number(s): |
|
33936 -- 1-hydroxy-naphthalene-3,6-disulfonic acid D3514 -- 4,4'-diisothio-cyanostilbene 2'2'-disulfonic acid |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1,000,000 units |
| |
|
|
|
Catalog Number: |
|
G5635 |
|
Activity: |
|
≥600 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more common: 67610, 73660, M1633 -- β-D-galactopyranoside 25589 -- β-D-lactoside 69601 -- β-D-galactopyranoside N1252 -- 4-Nitrophenyl β-D-galactopyranoside B4387 -- 5-Bromo-3-indolyl β-D-galactopyranoside 16665 -- 5-Bromo-4-chloro-3-indolyl β D-galactopyranoside F2756 -- Fluorescein di(galactopyranoside) |
|
Common Inhibitor (s) w Product Number(s): |
|
N2259 1-naphthyl-β-D-galactoside A9414, A6560, A9045, A0701 agarose * beta-galactopyranoside |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
4500 KU |
| |
|
|
|
Catalog Number: |
|
G0876 |
|
Activity: |
|
≥85,000 units/mL |
|
Common Substrate(s) w Product Number(s): |
|
69602 -- 4-Methylumbelliferyl-β-D-glucuronide hydrate N1875 -- Naphthol AS-BI β-D-glucuronide 77636 -- Phenolphthalein β-D-glucuronide R2886 -- Resorufin β-D-glucuronide sodium salt |
|
Common Inhibitor (s) w Product Number(s): |
|
T9165 ψ-Tectorigenin G7166 D-Glucaro-δ-lactam potassium salt solid G5269 D-Glucuronic acid |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
20,000 ml |
| |
|
|
|
Catalog Number: |
|
G7017 |
|
Activity: |
|
≥100,000 units/mL |
|
Common Substrate(s) w Product Number(s): |
|
69602 -- 4-Methylumbelliferyl-β-D-glucuronide hydrate N1875 -- Naphthol AS-BI β-D-glucuronide 77636 -- Phenolphthalein β-D-glucuronide R2886 -- Resorufin β-D-glucuronide sodium salt |
|
Common Inhibitor (s) w Product Number(s): |
|
T9165 -- ψ-Tectorigenin G7166 -- D-Glucaro-δ-lactam potassium salt solid G5269 -- D-Glucuronic acid |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
20,000 ml |
| |
|
|
|
Catalog Number: |
|
G8132 |
|
Activity: |
|
≥1,000,000 units/g solid |
|
Common Substrate(s) w Product Number(s): |
|
69602 -- 4-Methylumbelliferyl-β-D-glucuronide hydrate N1875 -- Naphthol AS-BI β-D-glucuronide 77636 -- Phenolphthalein β-D-glucuronide R2886 -- Resorufin β-D-glucuronide sodium salt |
|
Common Inhibitor (s) w Product Number(s): |
|
T9165 -- ψ-Tectorigenin G7166 -- D-Glucaro-δ-lactam potassium salt solid G5269 -- D-Glucuronic acid |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
800,000 KU |
| |
|
|
|
Catalog Number: |
|
C40 (C9631) |
|
Activity: |
|
≥10,000 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
Peroxide R425966 -- 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonic acid) D8001 -- 3,3'-diaminobenzidine A7631 -- ascorbate |
|
Common Inhibitor (s) w Product Number(s): |
|
A8056 -- 3-Amino-1,2,4-triazole M6250 -- 2-Mercaptoethanol |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
2,000 g |
| |
|
|
|
Catalog Number: |
|
DN25 (D0876) |
|
Activity: |
|
Protein ≥85 %, ≥400 Kunitz units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
A9272 3'AMP Double Stranded DNA circular plasmid DNA |
|
Common Inhibitor (s) w Product Number(s): |
|
P9375 1,10-Phenanthroline monohydrate E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid E0270 Ethylenediaminetetraacetic acid tripotassium salt dihydrate N9653 Netropsin dihydrochloride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
10,000 g |
| |
|
|
|
Catalog Number: |
|
D5025 |
|
Activity: |
|
Protein ≥80 %, ≥2,000 Kunitz units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
A9272 3'AMP Double Stranded DNA circular plasmid DNA |
|
Common Inhibitor (s) w Product Number(s): |
|
P9375 1,10-Phenanthroline monohydrate E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid E0270 Ethylenediaminetetraacetic acid tripotassium salt dihydrate N9653 Netropsin dihydrochloride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
600 g |
| |
|
|
|
Catalog Number: |
|
E3019 |
|
Activity: |
|
≥15 units/mg solid |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more common: N8505 1-Naphthyl acetate N8000 1-Naphthyl butyrate I9368 4-Nitrophenyl butyrate N0758 Naphthol AS-D chloroacetate 83636 Resorufin acetate |
|
Common Inhibitor (s) w Product Number(s): |
|
T4376 1-chloro-3-tosylamido-4-phenyl-2-butanone M6250 2-Mercaptoethanol 179124 Acetone 360457 Acetonitrile E8375 Eserine |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1,500,000 units |
| |
|
|
|
Catalog Number: |
|
F4165 |
|
Activity: |
|
≥0.1 unit/mg solid |
|
Common Substrate(s) w Product Number(s): |
|
C7078 Casein F5386 Fibrin |
|
Common Inhibitor (s) w Product Number(s): |
|
C8917 Cystatin from chicken egg white lyophilized powder E3132 E-64 |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1,300,000 units |
| |
|
|
|
Catalog Number: |
|
G4256 |
|
Activity: |
|
≥20 units/mg protein (modified Warburg-Christian) |
|
Common Substrate(s) w Product Number(s): |
|
G0380 alpha-D-galactose 1-phosphate U1756 UDP-glucose |
|
Common Inhibitor (s) w Product Number(s): |
|
131377 1,10-Phenanthroline 164984 8-Hydroxyquinoline A2754 ADP |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
15,000 units |
| |
|
|
|
Catalog Number: |
|
G6378 |
|
Activity: |
|
≥200 units/mg protein (modified Warburg-Christian) |
|
Common Substrate(s) w Product Number(s): |
|
09972 3-Nitrotetrazolium blue chloride 74030, 74032 Nitrotetrazolium blue chloride 88190 Tetrazolium blue chloride |
|
Common Inhibitor (s) w Product Number(s): |
|
A2056 Acetyl-CoA A2324, A3650 Amikacin A2754 ADP A5394 ATP N5130 NADPH |
|
Cofactor or Coenzyme: |
|
N0505 NADP |
|
Typical Batch Size: |
|
1,500,000 units |
| |
|
|
|
Catalog Number: |
|
I4504 |
|
Activity: |
|
>300 units/mg solid |
|
Common Substrate(s) w Product Number(s): |
|
72555 1-kestose 12255 inulin L8647 levane |
|
Common Inhibitor (s) w Product Number(s): |
|
M6250 2-Mercaptoethanol A6387 Ammonium sulfate A9880 Aniline A7506 Ascorbic acid |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
5000 g |
| |
|
|
|
Catalog Number: |
|
L0382 |
|
Activity: |
|
Minimum ≥20,000 units.mg protein (using olive oil) |
|
Common Substrate(s) w Product Number(s): |
|
61710 2-Naphthyl laurate 19362 4-Methylumbelliferyl butyrate T5069 4-Trifluoromethylumbelliferyl oleate 46942 Fluorescein dibutyrate 57420 Indoxyl acetate 83637 Resorufin butyrate |
|
Common Inhibitor (s) w Product Number(s): |
|
O4139 Orlistat |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
250,000, 000 units |
| |
|
|
|
Catalog Number: |
|
L9506 |
|
Activity: |
|
≥15 × 106 light units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
62645, L6882, 62643 D-Luciferin sodium salt L9504, L6152 D-Luciferin synthetic FFW Fireflies, whole F3641 Firefly Lantern powder FFT Firefly tails (lanterns) L9007 Luciferin 6'-ethyl ether L1147 Luciferin 6'-methyl ether |
|
Common Inhibitor (s) w Product Number(s): |
|
A9501, 851205 3',5'-cyclic AMP A6631 Arsenate 126799 Dodecanol E0270 EDTA |
|
Cofactor or Coenzyme: |
|
A5394 ATP |
|
Typical Batch Size: |
|
20,000 mg |
| |
|
|
|
Catalog Number: |
|
L7386 |
|
Activity: |
|
Protein ~60 % by biuret, ≥500 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
77140, 08678 Peptidoglycan E1625 Elastin |
|
Common Inhibitor (s) w Product Number(s): |
|
A6387, A4915 Ammonium Sulfate 131377 1,10-Phenanthroline A1593, A5354 Ampicillin E0270 EDTA I2512, I9148 Iodoacetate S9888, 310166 Sodium Chloride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
60 g |
| |
|
|
|
Catalog Number: |
|
L4402 |
|
Activity: |
|
Protein ~50 % by biuret, ≥2,000 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
77140, 08678 Peptidoglycan E1625 Elastin |
|
Common Inhibitor (s) w Product Number(s): |
|
A6387, A4915 Ammonium Sulfate 131377 1,10-Phenanthroline A1593, A5354 Ampicillin E0270 EDTA I2512, I9148 Iodoacetate S9888, 310166 Sodium Chloride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
3 g |
| |
|
|
|
Catalog Number: |
|
L6876 |
|
Activity: |
|
Protein ~95 %, ≥50,000 units/mg protein (E1%/282) |
|
Common Substrate(s) w Product Number(s): |
|
M5639 4-Methylumbelliferyl β-D-N,N′,N′′-triacetylchitotrioside N8638 4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose 77145 Peptidoglycan dyed with Remazol Brilliant Blue R 53243 Peptidoglycan from Micrococcus luteus |
|
Common Inhibitor (s) w Product Number(s): |
|
M6250, 63700 2-Mercaptoethanol Chitotetraose C1297, 451657 Copper Sulfate G7889 Glucosamine G4376 Glutathione H7125, 53290 Histamine P4707, P4832, P8920 Poly-L-lysine |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
30,000 g |
| |
|
|
|
Catalog Number: |
|
L2524 |
|
Activity: |
|
≥2,000 units/mg protein, Protein ≥20 % by biuret |
|
Common Substrate(s) w Product Number(s): |
|
51475 Yeast,G10120 Sephadex |
|
Common Inhibitor (s) w Product Number(s): |
|
M7504, M3640 Mannan |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
50,000,000 units |
| |
|
|
|
Catalog Number: |
|
M9901 |
|
Activity: |
|
≥4000 units/mg protein (biuret) |
|
Common Substrate(s) w Product Number(s): |
|
Streptococcus cell walls |
|
Common Inhibitor (s) w Product Number(s): |
|
18350, B81255 N-bromosuccinimide |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
100,000 KU |
| |
|
|
|
Catalog Number: |
|
N2876 |
|
Activity: |
|
Minimun 0.5 units.mg solid (using nan-lactose) Minumum 0.1 units/mg solid using bovine submaxillary mucin |
|
Common Substrate(s) w Product Number(s): |
|
M8639, 69587, N1516, B4666 ... A-D-N-acetylneuraminic acid |
|
Common Inhibitor (s) w Product Number(s): |
|
S8063 Siastatin B E0270 EDTA I1149,, I6125 Iodoacetamide M6250, 63700 Mercaptoethanol 18350, B81255 N-bromosuccinimide |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
190,000 units |
| |
|
|
|
Catalog Number: |
|
P6887 |
|
Activity: |
|
≥3,200 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
77431 Phe-Ala-Phe… 96150 Z-Glu-Tyr |
|
Common Inhibitor (s) w Product Number(s): |
|
P5318 Pepstatin A microbial P7626 Phenylmethanesulfonyl fluoride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
10,000 g |
| |
|
|
|
Catalog Number: |
|
P7012 |
|
Activity: |
|
≥2,500 units/mg protein (E1%/280) |
|
Common Substrate(s) w Product Number(s): |
|
77431 Phe-Ala-Phe… 96150 Z-Glu-Tyr |
|
Common Inhibitor (s) w Product Number(s): |
|
P5318 Pepstatin A microbial P7626 Phenylmethanesulfonyl fluoride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
10,000 g |
| |
|
|
|
Catalog Number: |
|
P6782 |
|
Activity: |
|
lyophilized powder, ≥1000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol) |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more popular 11565, A3219 2,2'-Azino bis… T8665, T0440, T0865 TMB liquid substrate T8768 TMB dihydrochloride hydrate -bulk D3939, D6815 DAB for immunohistology A4685, A8511 Luminol 74030, 74032 Nitrotetrazolium Blue chloride D4168, D4418, D4293 SIGMAFAST DMB P9187 SIGMAFAST OPD tablet D9154 o-Diansidine dihydrochloride 78449 o-phenylenediamine dihydrochloride |
|
Common Inhibitor (s) w Product Number(s): |
|
A5960, 95210 Ascorbic acid 318116 Diethyldithiocarbamate E0270 EDTA 55458, 467804 Hydroxylamine |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
80,000 mg |
| |
|
|
|
Catalog Number: |
|
P8250 |
|
Activity: |
|
≥150 units/mg solid (using pyrogallol) |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more popular 11565, A3219 2,2'-Azino bis… T8665, T0440, T0865 TMB liquid substrate T8768 TMB dihydrochloride hydrate -bulk D3939, D6815 DAB for immunohistology A4685, A8511 Luminol 74030, 74032 Nitrotetrazolium Blue chloride D4168, D4418, D4293 SIGMAFAST DMB P9187 SIGMAFAST OPD tablet D9154 o-Diansidine dihydrochloride 78449 o-phenylenediamine dihydrochloride |
|
Common Inhibitor (s) w Product Number(s): |
|
A5960, 95210 Ascorbic acid 318116 Diethyldithiocarbamate E0270 EDTA 55458, 467804 Hydroxylamine |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
250,000 KU |
| |
|
|
|
Catalog Number: |
|
P8375 |
|
Activity: |
|
≥250 units/mg solid (using pyrogallol) |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more popular 11565, A3219 2,2'-Azino bis… T8665, T0440, T0865 TMB liquid substrate T8768 TMB dihydrochloride hydrate -bulk D3939, D6815 DAB for immunohistology A4685, A8511 Luminol 74030, 74032 Nitrotetrazolium Blue chloride D4168, D4418, D4293 SIGMAFAST DMB P9187 SIGMAFAST OPD tablet D9154 o-Diansidine dihydrochloride 78449 o-phenylenediamine dihydrochloride |
|
Common Inhibitor (s) w Product Number(s): |
|
A5960, 95210 Ascorbic acid 318116 Diethyldithiocarbamate E0270 EDTA 55458, 467804 Hydroxylamine |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
250,000 KU |
| |
|
|
|
Catalog Number: |
|
P1146 |
|
Activity: |
|
≥3 units/mg solid |
|
Common Substrate(s) w Product Number(s): |
|
73736, 73737 4-nitrophenyl phosphate bis 57420 indoxyl acetate 72091 NBT-BCIP solution 70471, 70491, 70485, 70495, 70476, N6125, 70478 Naphthol AS-... 77630 Phenolphthalein bisphosphate pyridine 93380 Tris phosphate 49990 sn-Glycerol 3-posphate bis |
|
Common Inhibitor (s) w Product Number(s): |
|
A6631 Arsenate 277908 Ammonium molybdate C1297, 451657 Copper Sulfate E0270 EDTA 215309, 201154, 402931, 215384 Fluoride 243655, 308390 Molybdate |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1,000,000 units |
| |
|
|
|
Catalog Number: |
|
P1435 |
|
Activity: |
|
≥10 units/mg protein (modified Warburg-Christian) |
|
Common Substrate(s) w Product Number(s): |
|
73736, 73737 4-nitrophenyl phosphate bis 57420 indoxyl acetate 72091 NBT-BCIP solution 70471, 70491, 70485, 70495, 70476, N6125, 70478 Naphthol AS-... 77630 Phenolphthalein bisphosphate pyridine 93380 Tris phosphate 49990 sn-Glycerol 3-posphate bis |
|
Common Inhibitor (s) w Product Number(s): |
|
A6631 Arsenate 277908 Ammonium molybdate C1297, 451657 Copper Sulfate E0270 EDTA 215309, 201154, 402931, 215384 Fluoride 243655, 308390 Molybdate |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1,000,000 units |
| |
|
|
|
Catalog Number: |
|
P7923 |
|
Activity: |
|
≥4,000 DEA units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
Multiple substrates. Some of the popular are: N7255, 71105, N7000 1-naphtyl phosphate .. 09972 3-Nitrotetrazolium blue chloride M8883, M3168, M8168, 69605 4-methylumbelliferyl phosphate N3129, 73737, N3254 4-Nitophenyl phosphate B6149, 16676, B5667, 16672... 5-bromo-6-chloro-3-indolyl phosphate B1911, B3804 BCIP/NBT F8764 Fast red 70471, N2250, Naphthol AS |
|
Common Inhibitor (s) w Product Number(s): |
|
L9756 (−)-Tetramisole hydrochloride P9375 1,10-Phenanthroline monohydrate D6518 Diethylenetriaminepentaacetic acid E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid E0270 Ethylenediaminetetraacetic acid (EDTA) tripotassium salt dihydrate T1512 Tetramisole hydrochloride |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1,300,000 units |
| |
|
|
|
Catalog Number: |
|
P3397 |
|
Activity: |
|
≥100 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
G6750, G6875 alpha-D-glucose 1-phosphate G6526, G7879 Glucose 6-phosphate D6539 2-deoxyribose-1-phosphate |
|
Common Inhibitor (s) w Product Number(s): |
|
251275, C0759 Citrate C0759 EDTA 62360, 499811 Lithium |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
5,000,000 units |
| |
|
|
|
Catalog Number: |
|
P6621 |
|
Activity: |
|
≥200 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
73739, N5879 O-(4-Nitrophenylphosphoryl)choline |
|
Common Inhibitor (s) w Product Number(s): |
|
P9373 1,10-Phenanthroline monohydrate C2313 Compound 48/80 D6518 Diethylenetriaminepentaacetic acid E0270 EDTA |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
600,000 units |
| |
|
|
|
Catalog Number: |
|
P6635 |
|
Activity: |
|
≥20 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
10381 1,4-alpha-D-glucosyl 10118, A8515 Amylopectin 859656, A3508 Amylose G0885, 50571, G8876, 50576 Glycogen |
|
Common Inhibitor (s) w Product Number(s): |
|
C4642, C4767 Cyclodextrin F1502, F3627 D-Fructose 6-phosphate M2152 D-Mannose 1-phosphate |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
20,000 mg |
| |
|
|
|
Catalog Number: |
|
P6556 |
|
Activity: |
|
≥30 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
Naturally occuring & synthetic peptides A2714, A9329 Aldolase C7078, C3400, 22078 Casein |
|
Common Inhibitor (s) w Product Number(s): |
|
436143, 71728 SDS |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
450,000 mg |
| |
|
|
|
Catalog Number: |
|
P7768 |
|
Activity: |
|
≥350 units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
A2754 ADP 79415 Phosho(enol)pyruvic acidmonopotassium salt 79418 Phosphol(enol)pyruvic acid monosodium salt hydrate 79430 Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt 79435 Phospho(enol)putuvic acid trisodium salt heptahydrate |
|
Common Inhibitor (s) w Product Number(s): |
|
Q0125 Quercetin dihydrate |
|
Cofactor or Coenzyme: |
|
A2754..ADP or A5394..ATP |
|
Typical Batch Size: |
|
5000 KU |
| |
|
|
|
Catalog Number: |
|
R5000 |
|
Activity: |
|
≥50 Kunitz units/mg solid |
|
Common Substrate(s) w Product Number(s): |
|
83847 Ribonucleic acid |
|
Common Inhibitor (s) w Product Number(s): |
|
D5758 Diethyl pyrocarbonate R7253 Ribonuclease Inhibitor human lyophilized powder |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
500 g |
| |
|
|
|
Catalog Number: |
|
R5125 |
|
Activity: |
|
≥85 Kunitz units/mg protein |
|
Common Substrate(s) w Product Number(s): |
|
83847 Ribonucleic acid |
|
Common Inhibitor (s) w Product Number(s): |
|
D5758 Diethyl pyrocarbonate R7253 Ribonuclease Inhibitor human lyophilized powder |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
1500 g |
| |
|
|
|
Catalog Number: |
|
P5380 |
|
Activity: |
|
≥7 units/mg solid |
|
Common Substrate(s) w Product Number(s): |
|
C7078, C3400, 22078 Casein F5386 Fibrin |
|
Common Inhibitor (s) w Product Number(s): |
|
360457, A3396 Acetonitrile 24217, 105899 Cyclohexanol E0270 EDTA |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
500 g |
| |
|
|
|
Catalog Number: |
|
T3399 |
|
Activity: |
|
≥50 NIH units/mg protein (biuret) |
|
Common Substrate(s) w Product Number(s): |
|
T3068 ...chromogenic substrate 51010 ...hydrochloride hydrate 51015, B9385, B4028 ...hydrochloride T1637, 90175 ...p-nitroanilide acetate B7632, 13042 ...p-nitroanilide hydrochloride |
|
Common Inhibitor (s) w Product Number(s): |
|
D7910 3,4-Dichloroisocoumarin A8456 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride A6191 Antipain dihydrochloride from microbial source A9141 Antithrombin III from bovine plasma A2221 Antithrombin III from human plasma A6397 Antithrombin III from rat plasma D0879 Diisopropylfluorophosphate H7016, H7380 Hirudin from leeches E7888 N-Acetyl-eglin C recombinant, expressed in proprietary host |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
2050 KU |
| |
|
|
|
Catalog Number: |
|
T4648 |
|
Activity: |
|
≥40 NIH units/mg protein (biuret) |
|
Common Substrate(s) w Product Number(s): |
|
T3068 ...chromogenic substrate 51010 ...hydrochloride hydrate 51015, B9385, B4028 ...hydrochloride T1637, 90175 ...p-nitroanilide acetate B7632, 13042 ...p-nitroanilide hydrochloride |
|
Common Inhibitor (s) w Product Number(s): |
|
D7910 3,4-Dichloroisocoumarin A8456 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride A6191 Antipain dihydrochloride from microbial source A9141 Antithrombin III from bovine plasma A2221 Antithrombin III from human plasma A6397 Antithrombin III from rat plasma D0879 Diisopropylfluorophosphate H7016, H7380 Hirudin from leeches E7888 N-Acetyl-eglin C recombinant, expressed in proprietary host |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
200,000 KU |
| |
|
|
|
Catalog Number: |
|
T1063 |
|
Activity: |
|
≥2800 NIH units/mg protein (E1%/280, 18.3) |
|
Common Substrate(s) w Product Number(s): |
|
T3068 ...chromogenic substrate 51010 ...hydrochloride hydrate 51015, B9385, B4028 ...hydrochloride T1637, 90175 ...p-nitroanilide acetate B7632, 13042 ...p-nitroanilide hydrochloride |
|
Common Inhibitor (s) w Product Number(s): |
|
D7910 3,4-Dichloroisocoumarin A8456 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride A6191 Antipain dihydrochloride from microbial source A9141 Antithrombin III from bovine plasma A2221 Antithrombin III from human plasma A6397 Antithrombin III from rat plasma D0879 Diisopropylfluorophosphate H7016, H7380 Hirudin from leeches E7888 N-Acetyl-eglin C recombinant, expressed in proprietary host |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
900,000 KU |
| |
|
|
|
Catalog Number: |
|
T6632 |
|
Activity: |
|
≥900 units/mL |
|
Common Substrate(s) w Product Number(s): |
|
F0503, 46875 5-fluoro-2'-deoxyuridine B5002, 858811 bromodeoxyuridine 323616, 331902 deoxythymidine D5412, 31180 deoxyuridine T9250, 89270 thymidine U3750, U6381, 94320 uridine |
|
Common Inhibitor (s) w Product Number(s): |
|
B5002, 5-Bromo-2-deoxyuridine F0503, 46875 5-Fluorodeoxyuridine 18660, 852473 Bromouracil |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
30,000 KU |
| |
|
|
|
Catalog Number: |
|
T2807 |
|
Activity: |
|
≥500 units/mL |
|
Common Substrate(s) w Product Number(s): |
|
F0503, 46875 5-fluoro-2'-deoxyuridine B5002, 858811 bromodeoxyuridine 323616, 331902 deoxythymidine D5412, 31180 deoxyuridine T9250, 89270 thymidine U3750, U6381, 94320 uridine |
|
Common Inhibitor (s) w Product Number(s): |
|
B5002, 5-Bromo-2-deoxyuridine F0503, 46875 5-Fluorodeoxyuridine 18660, 852473 Bromouracil |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
30,000 KU |
| |
|
|
|
Catalog Number: |
|
94310 |
|
Activity: |
|
≥9 units/mg |
|
Common Substrate(s) w Product Number(s): |
|
U2875 -- Uric acid sodium salt |
|
Common Inhibitor (s) w Product Number(s): |
|
C1297 -- Copper Sulfate |
|
Cofactor or Coenzyme: |
|
|
|
Typical Batch Size: |
|
300 g |
| |
|
|
All products are in the PREMIUM level of the Enhanced Qualiity Profile except Proteinase K, catalog No. P2308, which is in the ELITE level
>> Questions? Inquiries? Contact a SAFC Supply Solutions Representative >> Go to the Enzyme Explorer @ SigmaAldrich.com >> Return to Enhanced Quality Profile Program Section |