Extranet 
Welcome to SAFC Supply Solutions
BIOEAZETM Single Use Disposables
Cell Culture
BioEaze LogoTransporting cell culture liquids effectively is an essential part of speed to market, making the choice of the right disposables system vital. BIOEAZE™ from SAFC is the all in one disposables solution.
Disposable technology provides many advantages, including the elimination of cleaning and sterilization procedures and the validation of these processes. SAFC provides standard and application specific systems, including the bioprocess bags, rigging sets, tubing, and other components. The bags, tubing and other components are available separately or assembled in total bioprocessing systems. BIOEAZE bioprocessing systems are available as sterile or non-sterile products.
SAFC can assemble a bioprocessing system based on your specific design or a representative from the SAFC bioprocess engineering team can meet with you to discuss your specific requirements, and then design a system to your exact needs.
>> As us about BIOEAZE™.
BIOEAZE™ Single Use Disposables
  Ethyl Vinyl Acetate bags
  Polyethylene Bags
  EZ Top and Asceptic Connecting Devices
  Rigging Sets
  Components
  Tank Liners
  Tubing
  Storage & Transport
BIOEAZE™ Bags - Ethyl Vinyl Acetate (EVA) Film (sterile)
Catalog # Name Product Overview
1522B BIOEAZE™ 350 mL Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 1 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Medicinal Port
· 1 Spike Port
Individually polybagged
pdf_icon11
1935B BIOEAZE™ 350 mL Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
1446B BIOEAZE™ 350 mL Ethyl Vinyl Acetate (EVA) Sterile Bag 10/pk
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 1.5 ft. length of 1/8" × 1/4" Clear 082 C-Flex® Tubing
· 1 Spike Port
· 1 Swabable Needleless Luer Valve
Individually polybagged
1521B BIOEAZE ™ 1 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for Terumo® Welding to 1331B, 1332B and 1333B BIOEAZE™ Polyethylene (PE) Sterile Bags
Designed with:
· 1 × 1 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Medicinal Port
· 1 Spike Port
Individually polybagged
1936B BIOEAZE™ 1 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
1937B BIOEAZE™ 2 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
1933B BIOEAZE™ 2 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
· 1 Male Luer Lock with Female Cap
· 1 Spike Port
Individually polybagged
1934B BIOEAZE™ 5 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for use with a Terumo® Aseptic Connecting Device
Designed with:
· 1 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
· 1 Male Luer Lock with Female Cap
· 1 Spike Port
Individually polybagged
1938B BIOEAZE™ 5 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
1523B BIOEAZE™ 5 - 10 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed with:
· 2 × 2.5 ft. length of Size 15 Opaque 001 C-Flex® Tubing
· 1/4" Male and Female Quick Connects with Caps
· 1 Spike Port
Double polybagged
1524B BIOEAZE™ 5 - 10 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 2 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Spike Port
Double polybagged
1329B BIOEAZE™ 10 - 20 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 2 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Spike Port
Double polybagged
1330B BIOEAZE™ 10 - 20 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed with:
· 2 × 2.5 ft. length of Size 15 Opaque 001 C-Flex® Tubing
· 1/4" Male and Female Quick Connects with Caps
· 1 Spike Port
Double polybagged
2339B BIOEAZE™ 500 L Polyethylene (PE) Sterile Bag
With 2 Top Discharge Ports Configured with:
· Port P1: Swabable Luer Lock
·
Port P2: 1 x 1 ft. length of 1/8” x 1/4” Clear 082 C-Flex® Tubing with Pinch Clamp and Plug
3392B BIOEAZE™ .5 - 1 L Ethyl Vinyl Acetate (EVA) Sterile Bag with BioQuate Disposable Aseptic Connector (DAC) Double Polybagged.
Configured with:
· Port P1: 1 x 6” length of 1/4” x 7/16” Clear 082 Clear C-Flex® Tubing with Pinch Clamp and Disposable Aseptic Connector (DAC)
· Port P2: 1 x 1 ft. length of Opaque 001 Size 16 C-Flex® Tubing with Pinch Clamp and Plug
·
Port P3: 1 Spike Port
Designed for use with the ReadyMate Disposable Aseptic Connector (DAC) or Terumo Aseptic Connecting Device (ACD)
  For ordering information please contact your local SAFC Supply Solutions office
back to top
BIOEAZE™ Bags - Polyethylene (PE) Film (sterile)
Catalog # Name Product Overview
1331B BIOEAZE™ 50 L Polyethylene (PE) Sterile Bag
Designed with:
· 1 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 2 × 2.5 ft. length of Size 73 Opaque 001 C-Flex® Tubing
· Male and Female Quick Connects with Caps
Double polybagged
1946B BIOEAZE™ 50 L Polyethylene (PE) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 2.5 ft. length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1 × 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
Double polybagged
1947B BIOEAZE™ 100 L Polyethylene (PE) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 2.5 ft. length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1 × 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
Double polybagged
1332B BIOEAZE™ 100 L Polyethylene (PE) Sterile Bag
Designed with:
· 1 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 2 × 2.5 ft. length of Size 73 Opaque 001 C-Flex® Tubing
· Male and Female Quick Connects with Caps
Double polybagged
1333B BIOEAZE™ 200 L Polyethylene (PE) Sterile Bag
Designed with:
· 1 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 2 × 2.5 ft. length of Size 73 Opaque 001 C-Flex® Tubing
· Male and Female Quick Connects with Caps
Double polybagged
1988B BIOEAZE™ 200 L Polyethylene (PE) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 2.5 ft. length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1 × 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
Double polybagged
1826B BIOEAZE™ 500 L Polyethylene (PE) Sterile Bag
Designed to fit 40.9" × 47.25" × 50.4" tote
Designed with:
· 3 × 10 ft. length of 3/8" × 5/8" Clear 082 C-Flex® Tubing
· Male and Female Quick Connects with Caps
· Bottom discharge consisting of 1 × 10 ft. length of 3/8" × 5/8" Clear 082 C-Flex® Tubing
Double polybagged
  For ordering information please contact your local SAFC Supply Solutions office
back to top
EZ Top and Asceptic Connecting Devices
Catalog # Name
2503S Fits 500 mL or 1 L Bottle with 082 C-Flex® Stopper and 38 mm Cap with 1 x 2 ft. length of 1/8” x 1/4” 082 Clear C-Flex® Tubing with PInch Clamp and Plug. Terumo application
2504S Fits 500 mL or 1 L Bottle with 374 (Animal-Free) C-Flex® Stopper and 38 mm Cap with 1 x 2 ft. length of 1/8” x 1/4” 374 Clear C-Flex® Tubing with PInch Clamp and Plug. Terumo application
2505S BIOEAZE™ 500 mL Bottle with 082 Clear C-Flex® Stopper and 38 mm Cap with 1 x 2 ft. length of 1/8” x 1/4” 082 Clear C-Flex® Tubing with PInch Clamp and Plug Terumo application
2502S Fits 500 mL or 1 L Bottle with 374 (Animal-Free) C-Flex® Stopper with 38 mm Cap with 1 x 2 ft. length of 3/16” x 3/8” 374 Clear C-Flex® Tubing with Pinch Clamp and 1/4” Female Polysulfone Quick Connect with Cap. Quick Connect application
2501S Fits 500 mL or 1 L Bottle with 082 Clear C-Flex® Stopper with 38 mm Cap with 1 x 2 ft. length of 3/16” x 3/8” Clear 082 C-Flex® Tubing with Pinch Clamp and 1/4” Female Polycarbonate Quick Connect with Cap. Quick Connect Application
3388P BIOEAZE™ 1/4” Barbed Disposable Aseptic Connector (DAC) 2/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3393P BIOEAZE™ 1/4” Barbed Disposable Aseptic Connector (DAC) 10/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3389P BIOEAZE™ 3/8” Barbed Disposable Aseptic Connector (DAC) 2/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3394P BIOEAZE™ 3/8” Barbed Disposable Aseptic Connector (DAC) 10/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3390P BIOEAZE™ 1/2” Barbed Disposable Aseptic Connector (DAC) 2/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3395P BIOEAZE™ 1/2” Barbed Disposable Aseptic Connector (DAC) 10/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3391P BIOEAZE™ 3/4” Barbed Disposable Aseptic Connector (DAC) 2/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
3396P BIOEAZE™ 3/4” Barbed Disposable Aseptic Connector (DAC) 10/pk (Autoclavable). Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
2508P BIOEAZE™ BioQuate Tamper Evident Zip Clamp Double Polybagged 1/pk. Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
2509P BIOEAZE™ BioQuate Tamper Evident Zip Clamp Double Polybagged 5/pk. Designed for use with the ReadyMate Disposable Asceptic Connector or the Terumo Aseptic Connecting device
  For ordering information please contact your local SAFC Supply Solutions office
back to top
BIOEAZE™ Rigging Sets (sterile)
Catalog # Name Product Overview
1336R BIOEAZE™ Rigging Set 1/4" with Male-Female Quick Connects Sterile
C-Flex® Tubing, Size 15 Opaque 001
· with a 1/4" Male Quick Connect
· with a Cap at one end and a Female Quick Connect at the other end
Individually double polybagged
10 ft. length
1337R BIOEAZE™ Rigging Set 3/8" with Male-Female Quick Connects Sterile
C-Flex® Tubing, Size 73 Opaque 001
· with a 3/8" Male Quick Connect
· with a Cap at one end and a Female Quick Connect at the other end
Individually double polybagged
10 ft. length
1520R BIOEAZE™ Rigging Set 3/8" with Male-Female Quick Connects Sterile
C-Flex® Tubing, Size 73 Opaque 001
· with a 3/8" Male Quick Connect
· with a Cap at one end and a Female Quick Connect at the other end
Individually double polybagged
25 ft. length
1339R BIOEAZE™ Rigging Set Aseptic Connecting Device (ACD) Clear Sterile
C-Flex® Tubing, 1/8" x 1/4" Clear 082-C
· for use with Aseptic Connecting Device (ACD)
· with plugs at each end
Individually double polybagged
10 ft. length
1338R BIOEAZE™ Rigging Set Aseptic Connecting Device (ACD) Sterile
C-Flex® Tubing, Size 16 Opaque 001
· for use with Aseptic Connecting Device (ACD)
· with plugs at each end
Individually double polybagged
10 ft. length
1654R BIOEAZE™ Dispensing Tube Set Sterile
· with Male Quick Connect
Individually double polybagged
2 ft. length
1655R BIOEAZE™ Dispensing Tube Set Sterile
· with Aseptic Connecting Device (ACD) Weldable Size 16 Opaque 001 C-Flex® Tubing
Individually double polybagged
5 ft. length
1656R BIOEAZE™ Dispensing Tube Set Sterile
· with Aseptic Connecting Device (ACD) Weldable 1/8" x 1/4" Clear 082 C-Flex® Tubing
Individually double polybagged
5 ft. length
3677R BIOEAZE™ Rigging Set Sterile. Designed for use with Terumo® Aseptic Connecting Device (ACD) Applications, Double Polybagged
· 5 x 1 ft. lengths of 1/8” x 1/4” Clear 082 C-Flex® Tubing with Plugged Ends 5/pk
3678R BIOEAZE™ Rigging Set Sterile. Designed for use with Terumo® Aseptic Connecting Device (ACD) Applications, Double Polybagged
· 5 x 1 ft. lengths of Size 16 Opaque 001 C-Flex® Tubing with Plugged Ends 5/pk
2671R BIOEAZE™ Rigging Set Sterile. Designed for use with the SAFC Biosciences’ QUICK SCALE™ Program to Allow for Multiple Applications, Double Polybagged.
· 3/16” x 3/8” Silicone Tubing with Female Quick Connect on One End and Filling Bell with Cap on One End
2672R BIOEAZE™ Rigging Set Sterile. Designed for use with the SAFC Biosciences’ QUICK SCALE™ Program to Allow for Multiple Applications, Double Polybagged.
· 3/8” x 5/8” Clear C-Flex® Tubing with Female Quick Connect on One End and Colder Mini-Sanitary Steam-Thru Connector on One End
2674R BIOEAZE™ Rigging Set Sterile. Designed for use with the SAFC Biosciences’ QUICK SCALE™ Program to Allow for Multiple Applications, Double Polybagged.
· 1/8” x 1/4” Clear C-Flex® Tubing on One End and Female Quick Connect on One End
2675R BIOEAZE™ Rigging Set Sterile. Designed for use with the SAFC Biosciences’ QUICK SCALE™ Program to Allow for Multiple Applications, Double Polybagged.
· Female Quick Connect on One End and Female Quick Connect and Female MPX (1/2”) Quick Connect on One End
2677R BIOEAZE™ Rigging Set Sterile. Designed for use with the SAFC Biosciences’ QUICK SCALE™ Program to Allow for Multiple Applications, Double Polybagged.
· Female Quick Connect on One End with 3/4” Sanitary Fitting on One End and 1.5” Sanitary Fitting on One End
1814R BIOEAZE™ Syringe Sampling Set Sterile - Designed for Sampling with Terumo® Aseptic Cennecting Device (ACD) Applications
· 1.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing with 30 cc and 60 cc syringes
  For ordering information please contact your local SAFC Supply Solutions office
back to top
BIOEAZE™ Components
Catalog # Name Product Overview
1507P BIOEAZE™ Male Quick Connect Polycarbonate 1/4" 25/pk
1514P BIOEAZE™ Female Quick Connect Polycarbonate 1/4" 25/pk
1508P BIOEAZE™ Male Quick Connect Polycarbonate 3/8" 25/pk
1515P BIOEAZE™ Female Quick Connect Polycarbonate 3/8" 25/pk
1509P BIOEAZE™ Female Quick Cap Polycarbonate 25/pk
1516P BIOEAZE™ Male Quick Cap Polycarbonate 25/pk
1512P BIOEAZE™ Male Luer Lock Polycarbonate 1/4" 50/pk
1504P BIOEAZE™ Female Luer Lock Polycarbonate 1/4" 50/pk
1511P BIOEAZE™ Male Luer Lock Polycarbonate 3/16" 50/pk
1506P BIOEAZE™ Female Luer Lock Polycarbonate 3/16" 50/pk
1505P BIOEAZE™ Male Luer Cap Polycarbonate 50/pk
1513P BIOEAZE™ Female Luer Cap Polycarbonate 50/pk
1510P BIOEAZE™ Mini Sanitary Fitting, Tri-Clamp Connector with 3/8" Barb 25/pk
1801P BIOEAZE™ Mini Sanitary Fitting, Tri-Clamp Connector with 1/8" Barb 25/pk
1802P BIOEAZE™ Mini Sanitary Fitting, Tri-Clamp Connector with 1/4" Barb 25/pk
1803P BIOEAZE™ Mini Sanitary Fitting, Tri-Clamp Connector with 1/2" Barb 25/pk
1804P BIOEAZE™ Mini Sanitary End Cap 25/pk
1805P BIOEAZE™ Mini Sanitary Push/Pull Clamp 25/pk
1806P BIOEAZE™ Sanitary 1" Tri-Clamp Connector with 1/4" Barb 25/pk
1807P BIOEAZE™ Sanitary 1" Tri-Clamp Connector with 3/8" Barb 25/pk
1808P BIOEAZE™ Sanitary 1" Tri-Clamp Connector with 1/2" Barb 25/pk
1809P BIOEAZE™ Sanitary 1" Tri-Clamp Connector with 3/4" Barb 25/pk
2615P BIOEAZE™ Male Quick Connect Polycarbonate 1/2” 25/pk (Autoclavable)
3204P BIOEAZE™ Female Quick Connect Polycarbonate 1/2” 25/pk (Autoclavable)
2617P BIOEAZE™ Male Quick Cap Polycarbonate for 1/2” Female Quick Connect 25/pk (Autoclavable)
2616P BIOEAZE™ Female Quick Cap Polycarbonate for 1/2” Male Quick Connect 25/pk (Autoclavable)
2185P BIOEAZE™ Male Quick Cap Polycarbonate for 1/4” and 3/8” Female Quick Connect 10/pk (Individually Bagged) Sterile
3654P BIOEAZE™ Female Quick Cap Polycarbonate for 1/4” and 3/8” Male Quick Connect 10/pk (Individually Bagged) Sterile
  For ordering information please contact your local SAFC Supply Solutions office
back to top
BIOEAZE™ Tank Liners (sterile)
Catalog # Name Product Overview
1569L BIOEAZE™ Tank Liner 50 L, PL-1026 20/cs
1570L BIOEAZE™ Tank Liner 130 L, PL-1026 20/cs
1571L BIOEAZE™ Tank Liner 200 L, PL-1026 20/cs
2220L BIOEAZE™ Tank Liner 50 L, 13.5” diameter x 29” height 20/pk. "3D" Style
2221L BIOEAZE™ Tank Liner 100 L, 18.5” diameter x 33” height 20/pk. "3D" Style
2222L BIOEAZE™ Tank Liner 200 L, 22.5” diameter x 38” height 20/pk. "3D" Style
  For ordering information please contact your local SAFC Supply Solutions office
back to top
BIOEAZE™ Tubing
Catalog # Name Product Overview
1581T BIOEAZE™ Tubing C-Flex®, Opaque Size 16 50 ft. coil
1582T BIOEAZE™ Tubing C-Flex®, Opaque Size 15 50 ft. coil
1583T BIOEAZE™ Tubing C-Flex®, Opaque Size 73 50 ft. coil
1584T BIOEAZE™ Tubing C-Flex®, Opaque Size 82 50 ft. coil
1585T BIOEAZE™ Tubing Platinum Cured Silicone 1/4" x 3/8" 50 ft. coil
1586T BIOEAZE™ Tubing Platinum Cured Silicone 3/8" x 5/8" 50 ft. coil
1587T BIOEAZE™ Tubing Platinum Cured Silicone 1/2" x 3/4" 50 ft. coil
  For ordering information please contact your local SAFC Supply Solutions office
back to top
BIOEAZE™ Storage and Transport
Catalog # Name Product Overview
89301 BIOEAZE™ Standard Drum Dolly
1 each
89303 BIOEAZE™ Drum 20 Gallon/50 L Straight Side, with lid
1 each
89304 BIOEAZE™ Drum 35 Gallon/100 L Straight Side, with lid
1 each
89305 BIOEAZE™ Drum 55 Gallon/200 L Straight Side, with lid
1 each
89306 BIOEAZE™ Plastic Tote, 21" x 15" x 9" Stackable
1 each
89310 BIOEAZE™ Arca ComboLife 285 Bulk Container
1 each
  For ordering information please contact your local SAFC Supply Solutions office
back to top