Extranet 
Welcome to SAFC Supply Solutions
Media
Cell Culture
SAFC is the largest supplier of dry powder media in the world. The company is a class 1 Medial Device manufacturer, subject to inspections by the FDA and maintains quality standards consistent with the Quality System Requirements, including Design Controls, as outlined in 21CFR Part 820.
All large-scale media and salt mixtures are produced in accordance with cGMPs and are suitable for manufacturing and research applications.
Chemicals used in the manufacture of cell culture media and salt mixtures conform, where applicable, to the published standards of the United States Pharmacopeia (USP), American Chemical Society (ACS), National Formulary (NF) or Food Chemical Codex (FCC). Chemicals meeting European Pharmacopoeia (EP) are available upon request.
Most media formulations contain amino acid and vitamin components that are manufactured by fermentation. None of these products are derived from animal products. For several amino acids, small quantities of milk casein hydrolysates may be used in the early laboratory seed stages as an essential nitrogen source. Also, milk derivatives are generally used in the industry for the cryopreservation of microorganisms. Porcine enzymes from Japan or plant enzymes are used during the digestion process of milk casein. SAFC has confirmed that casein hydrolysates are derived from the milk of dairy cattle from Australia or New Zealand, USDA regulated and approved sources determined to be free of BSE. The milk is obtained from healthy animals and collected under the same conditions as those required for human consumption. Some cultures may contain fish meat extract from Bonito sourced from Japan.
All dry powder media and salts are manufactured in environments controlled for relative humidity and temperature. All water used in the manufacture of liquid media meets the criteria as specified in the USP and EUP monographs for Water for Injection.
>> As us about Quick Scale bulk cGMP liquid media for batch sizes greater than 200L.
Classical Media:
  Mammalian Media
  Insect Media
Mammalian Media
Catalog # Name
B1522 Basal Medium Eagle
With Earle's salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested
51800C Claycomb Medium
without L-glutamine, sterile-filtered
D5523 Dulbecco's Modified Eagle's Medium/Low Modified
With 1000 mg/L dextrose, with 4.0 mM L-glutamine, with 110 mg/L sodium pyruvate, without sodium bicarbonate, dry powder, cell culture tested
51441C Dulbecco's Modified Eagle's Medium/High Modified
liquid
D5648 Dulbecco’s Modified Eagle’s Medium - high glucose
With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate., powder, cell culture tested
56436C Dulbecco's Modified Eagle's Medium/High Modified
dry powder, with 4500 mg/L dextrose, with 4.0 mM L-glutamine, without phenol red, without sodium bicarbonate, without sodium pyruvate, cell culture tested
D7777 Dulbecco’s Modified Eagle’s Medium - high glucose
With 4500 mg/L glucose, L-glutamine, and sodium pyruvate, without sodium bicarbonate., powder, cell culture tested
51449C Dulbecco's Modified Eagle's Medium/High Modified
with 4500 mg/L dextrose, with 110mg/L sodium pyruvate, without L-glutamine, without phenol red, liquid
51448C Dulbecco's Modified Eagle's Medium/Ham's Nutrient Mixture F12
cell culture tested
56498C Dulbecco's Modified Eagle's Medium/Ham's Nutrient Mixture F12
powder, with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 55 mg/L sodium pyruvate, without sodium bicarbonate
51445C Dulbecco's Modified Eagle's Medium/Ham's Nutrient Mixture F12
liquid, cell culture tested
D8900 Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham
With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, cell culture tested
51435C Dulbecco’s Modified Eagle’s Medium/High Modified
51492C Glasgow's Modified Eagle's Medium
I3390 Iscove’s Modified Dulbecco’s Medium
With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested, hybridoma tested
51651C Ham's Nutrient Mixture F12
liquid
N6760 Nutrient Mixture F-12 Ham
With L-glutamine, without sodium bicarbonate, powder, cell culture tested
I7633 Iscove’s Modified Dulbecco’s Medium
With L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, cell culture tested
56312C Medium 199 Modified
with Earle's Balanced Salts, with 0.68 mM L-glutamine, without sodium bicarbonate, dry powder
51322C Medium 199 Modified
liquid
51411C Minimum Essential Medium
liquid
56419C Minimum Essential Medium
with Earle's Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder
51412C Minimum Essential Medium
with Earle's Balanced Salts, without L-glutamine, liquid
51414C Minimum Essential Medium
liquid, cell culture tested
51415C Minimum Essential Medium
liquid
51416C Minimum Essential Medium
liquid, cell culture tested
56416C Minimum Essential Medium
with Earle's Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, cell culture tested
51417C Minimum Essential Medium
liquid, cell culture tested
M0894 Minimum Essential Medium Eagle
Alpha Modification, with L-glutamine, without ribonucleosides, deoxyribonucleosides and sodium bicarbonate, powder, cell culture tested
56449C Minimum Essential Medium Joklik's Modified
dry powder, with Earle's Balanced Salts, with 2.0 mM L-glutamine, without calcium chloride, without sodium bicarbonate
M0769 Minimum Essential Medium Eagle, Auto-Mod™
AutoMod™, with Earle's salts, without L-glutamine and sodium bicarbonate, powder, cell culture tested
56509C RPMI-1640 Medium
with 2.05 mM L-glutamine, without sodium bicarbonate, dry powder, cell culture test
51536C RPMI-1640 Medium
with 2.05 mM L-glutamine, with 25mM HEPES, liquid
Insect Media
Catalog # Name
T3285 TNM-FH insect medium
With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, insect cell culture tes
For ordering information please contact your local SAFC Supply Solutions office
back to top