Extranet 
Welcome to SAFC Supply Solutions
Sera
Cell Culture
All sera that are sourced from abattoirs in the US are collected at USDA inspected faculties. SAFC performs qualifying and routine audits at serum collection and processing sites. Donor sera are collected from monitored herds, maintained under veterinary care, and deemed healthy and suitable for human consumption. Australian and New Zealand blood product collections are done in AQIS and MAF inspected and qualified facilities. Both Australia and New Zealand are free from Bovine Spongifrom Encephalopathy (BSE) and other diseases such as Foot & Mouth Disease and Rinderpest.
Blood is aseptically collected under controlled conditions. Raw serum is immediately pooled and frozen. Thawed serum is filtered thru a series of membranes. Fetal Bovine Serum is filtered thru three 0.1micron filters and pooled All other sera are filtered thru three 0.2 micron filters. Sera are shipped frozen via air in insulated containers with dry ice.
A Certificate of Analysis indicating the country of origin is prepared for each lot of serum. SAFC guarantees traceability of serum materials to the point(s) of collection and the dates the whole blood was collected. The list of abattoirs, their identification numbers and collections dates are reviewed by Quality Assurance and retained with the batch record; which can be made available upon customer request.
>> Ask us about serum sample evaluation and lot reservations.
Fetal Bovine Serum:
  Fetal Bovine Serum - Sourced in the USA
  Fetal Bovine Serum - USDA Approved Source
  Fetal Bovine Serum - Sourced in Australia
  Fetal Bovine Serum - Sourced in New Zealand
Other Serum:
  Chicken Serum
  Goat Serum - Sourced in the USA
  Horse Serum - Sourced in the USA
  Human Serum
  Porcine Serum - Sourced in the USA
  Sheep Serum - Sourced in the USA
Bovine Serum:
  Adult Bovine Serum - Sourced in the USA
  Bovine Calf Serum - Sourced in the USA
  Bovine Calf Serum - Sourced in the Australia
  Newborn Calf Serum - Sourced in the USA
  Newborn Calf Serum - Sourced in New Zealand
Fetal Bovine Serum - Sourced in USDA Approved Countries
Catalog # Name
12103C Fetal bovine Serum –frozen
12106C Fetal Bovine Serum-heat inactivated & frozen
12107C Fetal Bovine Serum-frozen & gamma irradiated by SER-TAIN™ Process
12117C Fetal Bovine Serum-dialyzed, frozen and gamma irradiated by SER-TAIN™ Process
12176C Fetal Bovine Serum-heat inactivated, frozen & gamma irradiated by SER-TAIN™ Process
Fetal Bovine Serum - Sourced in USDA
Catalog # Name
12303C Fetal Bovine Serum-frozen
12306C Fetal bovine Serum-heat inactivated & frozen
12307C Fetal Bovine Serum-frozen & gamma irradiated by SER-TAIN™ Process
Fetal Bovine Serum - Sourced in Australia
Catalog # Name
12003C Fetal Bovine Serum-frozen
12006C Fetal bovine Serum-heat inactivated & frozen
12007C Fetal Bovine Serum-frozen & gamma irradiated by SER-TAIN™ Process
Fetal Bovine Serum - Sourced in New Zealand
Catalog # Name
12203C Fetal Bovine Serum-frozen
12206C Fetal bovine Serum-heat inactivated & frozen
12207C Fetal Bovine Serum-frozen & gamma irradiated by SER-TAIN™ Process
For ordering information please contact your local SAFC Supply Solutions office
back to top
Adult Bovine Serum - Sourced in USA
Catalog # Name
12153C Adult Bovine Serum-sourced in the USA-frozen
12157C Fetal Bovine Serum-frozen & gamma irradiated by SER-TAIN™ Process
Bovine Calf Serum - Sourced in USA & Australia
Catalog # Name
12133C Bovine Calf Serum-sourced in the USA-frozen
12136C Bovine Calf Serum-sourced in the USA-heat inactivated & frozen
12137C Bovine Calf Serum-sourced in the USA-gamma irradiated by SER-TAIN™ Process & frozen
12138C Bovine Calf Serum, sourced in the USA-iron fortified & frozen
12238C Bovine Calf Serum-sourced in the USA-iron fortified, gamma irradiated by the SER-TAIN™ Process & frozen
12043C Bovine Calf Serum-donor sourced in Australia-frozen
12046C Bovine Calf Serum-donor sourced in Australia-heat inactivated & frozen
12047C Bovine Calf Serum-donor sourced in Australia-gamma irradiated by SER-TAIN™ Process & frozen
13063C Bovine Calf Serum-sourced in Australia-frozen
Newborn Calf Serum - Sourced in USA & New Zealand
Catalog # Name
12123C Newborn Bovine Serum-sourced in the USA-frozen
12126C Newborn Bovine Serum-sourced in the USA-heat inactivated & frozen
12127C Newborn Bovine Serum-sourced in the USA- gamma irradiated by SER-TAIN™ & frozen
12023C Newborn bovine Serum-sourced in New Zealand-frozen
12026C Newborn Calf Serum-sourced in New Zealand-heat inactivated & frozen
12027C Newborn Calf Serum-sourced in New Zealand-gamma irradiated by SER-TAIN™ Process & frozen
For ordering information please contact your local SAFC Supply Solutions office
back to top
Chicken Serum - Sourced in USA
Catalog # Name
C5405 Chicken Serum
Goat Serum - Sourced in USA
Catalog # Name
12031C Goat Serum- donor sourced in the USA, frozen
12036C Goat Serum- donor sourced in the USA, heat inactivated & frozen
12037C Goat Serum- donor sourced in the USA, gamma irradiated by SER-TAIN™ Process & frozen
Horse Serum - Sourced in USA
Catalog # Name
12449C Horse Serum- donor sourced in the USA, frozen
12446C Horse Serum- donor sourced in the USA, heat inactivated & frozen
12447C Horse Serum- donor sourced in the USA, gamma irradiated by SER-TAIN™ Process & frozen
Human Serum
Catalog # Name
H4522 Human Serum - from human male AB plasma, sterile-filtered
S7023 Human Serum
Porcine Serum - Sourced in USA
Catalog # Name
12731C Porcine Serum-sourced in the USA, frozen
12736C Porcine Serum-sourced in the USA-heat inactivated & frozen
12737C Porcine Serum- sourced in the USA, gamma irradiated by SER-TAIN™ Process & frozen
Sheep Serum - Sourced in USA
Catalog # Name
S2263 Sheep Serum-sourced in the USA, collected from adult animals, frozen
For ordering information please contact your local SAFC Supply Solutions office
back to top