Extranet 
Welcome to SAFC Supply Solutions
Proteins & Enzymes
Diagnostics Product Group
 On this Page
Enzymes and Inhibitors
Nucleosides, Nucleotides, and Cofactors
Proteins and Derivatives
Substrates
Table bottom
Enzymes and Inhibitors
Catalog # Name
A7011 Alcohol Dehydrogenase Saccharomyces cerevisiae
Lyophilized powder (contains buffer salts), activity: ≥300 units/mg protein
A8435 Alcohol Dehydrogenase, NADP+ dependent Thermoanaerobium brockii
Lyophilized powder, activity: 5-15 units/mg protein
A0438 Alcohol Oxidase from Hansenula sp.
Vacuum-dried powder, activity: 20-40 units/mg protein (Bradford)
A8811 Aldolase from rabbit muscle
Ammonium sulfate suspension, activity: 10-20 units/mg protein
A2714 Aldolase from rabbit muscle
Lyophilized powder, activity: ~10 units/mg protein
A1603 Alginate Lyase from Flavobacterium sp.
A8003 Allopurinol
A6664 4-Amidinophenylmethanesulfonyl fluoride hydrochloride
A4257 L-Amino Acid Oxidase Bothrops atrox venom
Type II, dried venom, activity: ≥0.2 unit/mg solid
A8200 Aminopeptidase from Aeromonas proteolytica
Lyophilized powder, activity: 50-150 units/mg protein
A3403 α-Amylase
Type XII-A, saline solution, activity: 500-1,000 units/mg protein (biuret)
A9972 α-Amylase
Lyophilized powder, activity: ≥100 units/mg protein
A3176 α-Amylase
Type VI-B, activity: 10-30 units/mg solid
A4268 α-Amylase
Type I-A, PMSF treated, saline suspension, activity: 700-1400 units/mg protein (E1%/280)
A2330 Amyloglucosidase from Candida tsukubaensis
Ammonium sulfate suspension, activity: 50-150 units/mg protein
A9285 alpha;1-Antichymotrypsin from human plasma
~95% (SDS-PAGE), lyophilized powder
A9024 alpha;1-Antitrypsin from human plasma
Salt-free, lyophilized powder
A6150 alpha;1-Antitrypsin from human plasma
Salt-free, lyophilized powder
A4529 Aprotinin bovine lung
A6012 Aprotinin bovine lung
A6103 Aprotinin bovine lung
Recombinant, expressed in, activity: >5 TIU/mg protein
A0157 Ascorbate Oxidase Cucurbita sp.
Lyophilized powder, activity: 1,000-3,000 units/mg protein
B8385 Bestatin hydrochloride
≥98% (HPLC)
C1057 Butyrylcholinesterase equine serum
Lyophilized powder, activity: ≥900 units/mg protein
C3934 Carbonic Anhydrase bovine erythrocytes
Lyophilized powder, activity: ≥2,500 W-A units/mg protein
C2273 Carbonic Anhydrase bovine erythrocytes
For use as a marker in SDS-PAGE
C4396 Carbonic Anhydrase I human erythrocytes
Lyophilized powder, activity: 100-500 W-A units/mg protein
C3640 Carbonic Anhydrase Isozyme II from bovine erythrocytes
Lyophilized powder, activity: ≥2,000 W-A units/mg protein
C2522 Carbonic Anhydrase Isozyme II from bovine erythrocytes
Lyophilized powder, activity: ≥3,000 W-A units/mg protein
C9584 Carboxypeptidase B porcine pancreas
Lyophilized powder
C4899 Carnitine Acetyltransferase pigeon breast muscle
Ammonium sulfate suspension, activity: ≥80 units/mg protein
C3515 Catalase
Ammonium sulfate suspension, activity: 4,000-8,000 units/mg protein
C9322 Catalase
Lyophilized powder, activity: 2,000-5,000 units/mg protein
C30 Catalase
Aqueous suspension, activity: 10,000-40,000 units/mg protein
C40 Catalase
Lyophilized powder, activity: ≥10,000 units/mg protein
C100 Catalase
Aqueous suspension, activity: 40,000-60,000 units/mg protein (E1%/405)
C0150 Cathepsin B from human placenta
Lyophilized powder, activity: ≥5 units/mg protein
C9033 beta;-Chloro-L-alanine hydrochloride
C4911 Chlorothiazide
C3766 Cholesterol Esterase bovine pancreas
Lyophilized powder, activity: ≥200 units/g protein
C5896 Choline Oxidase from Alcaligenes sp.
Lyophilized powder, activity: ~10 units/mg solid
C7762 α-Chymotrypsin
Type I-S, essentially salt-free, lyophilized powder
C6423 α-Chymotrypsin
Suitable for protein sequencing, salt-free, lyophilized powder
C8946 α-Chymotrypsin
Lyophilized powder
C3142 alpha;-Chymotrypsin TLCK Treated from bovine pancreas
Type VII, essentially salt-free, lyophilized powder, activity: ≥40 units/mg protein
C7926 Collagenase from Clostridium histolyticum
C3921 Creatinase Pseudomonas sp.
Recombinant, expressed in, lyophilized powder, activity: 10-15 units/mg protein
C0984 Creatine Kinase MB Fraction from human heart
~700 I.U./mL, >90% (SDS-PAGE), liquid
C7886 Creatine Phosphokinase
Type III, salt-free, lyophilized powder, activity: ≥50 units/mg protein
C6638 Creatine Phosphokinase
Type IV, lyophilized powder, activity: 50-150 units/mg protein
C3755 Creatine Phosphokinase
Type I, salt-free, lyophilized powder, activity: 150-350 units/mg protein
C3172 Creatininase Pseudomonas sp.
Lyophilized powder, activity: 100-300 units/mg protein
C0733 Cyclooxygenase 1 from sheep
Aqueous solution, ≥95% (SDS-PAGE), activity: ≥40,000 units/mg protein
C0858 Cyclooxygenase 2 human
≥70% (SDS-PAGE), recombinant, expressed in, aqueous solution, activity: ≥8000 units/mg protein
C3805 Cyclophilin A human
>95% (SDS-PAGE), recombinant, expressed in, buffered aqueous solution
D5025 Deoxyribonuclease I from bovine pancreas
Type IV, lyophilized powder, Protein: ~90%, activity: ≥2,000 Kunitz units/mg protein
D4527 Deoxyribonuclease I from bovine pancreas
Type II, lyophilized powder, Protein: ~90%, activity: ≥2,000 units/mg protein
D4513 Deoxyribonuclease I from bovine pancreas
Type II-S, lyophilized powder, Protein: ~90%, activity: ≥2,000 units/mg protein
D4263 Deoxyribonuclease I from bovine pancreas
Standardized vial containing 2,000 Kunitz units of DNase I (D4527).
D1501 Deoxyribonucleic acid sodium salt from calf thymus
Type I, fibers
D4022 P1,P5-Di(adenosine-5′) pentaphosphate pentasodium salt
≥95% (HPLC), powder
D1387 P1,P3-Di(adenosine-5′) triphosphate ammonium salt
D1762 P1,P5-Di(guanosine-5′) pentaphosphate ammonium salt
D1028 P1,P4-Di(guanosine-5′) tetraphosphate ammonium salt
~95% (HPLC)
D2012 P1,P3-Di(guanosine-5′) triphosphate ammonium salt
≥90% (HPLC)
G4256 Galactose-1-phosphate Uridyltransferase from galactose-adapted yeast
Type IV, lyophilized powder, activity: 20-60 units/mg protein (modified Warburg-Christian)
G6125 Glucose Oxidase Aspergillus niger
Type II, activity: 15,000-25,000 units/g solid (without added oxygen)
G6378 Glucose-6-phosphate Dehydrogenase
Type XV, lyophilized powder, activity: 200-400 units/mg protein (modified Warburg-Christian)
G7877 Glucose-6-phosphate Dehydrogenase
Type VII, ammonium sulfate suspension, activity: ≥200 units/mg protein
G7750 Glucose-6-phosphate Dehydrogenase
Type V, dry ammonium sulfate cake, activity: ≥150 units/mg protein (modified Warburg-Christian)
G8404 Glucose-6-phosphate Dehydrogenase
Recombinant, expressed in, ammonium sulfate suspension, activity: 550-1,100 units/mg protein (biuret)
G8529 Glucose-6-phosphate Dehydrogenase
Recombinant, expressed in, lyophilized powder, activity: 550-1,100 units/mg protein (biuret)
G3651 alpha;-Glucosidase from Bacillus stearothermophilus
Lyophilized powder, activity: ≥50 units/mg protein
G7396 β-Glucuronidase
Type IX-A, lyophilized powder, activity: 1,000,000-5,000,000 units/g protein (30 min assay)
G7896 β-Glucuronidase
Type X-A, lyophilized powder, activity: 20,000,000-60,000,000 units/g protein, pH 6.8 (30 min assay)
G4259 β-Glucuronidase
Type HA-4, activity: 250,000-500,000 units/g solid
G0876 β-Glucuronidase
Type H-2, aqueous solution, ≥85,000 units/mL
G8132 β-Glucuronidase
Type L-II, lyophilized powder, activity: 1,000,000-3,000,000 units/g solid
G7005 Glutamic-Oxalacetic Transaminase from porcine heart
Type II-A, lyophilized powder, activity: 100-400 units/mg protein
G8255 Glutamic-Pyruvic Transaminase from porcine heart
Lyophilized powder, activity: ≥75 units/mg protein
G2267 Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle
Lyophilized powder, activity: ≥75 units/mg protein
G1642 sn-Glycerol-3-phosphocholine Phosphodiesterase mold
Lyophilized powder, activity: 5-20 units/mg protein
H4502 Hexokinase from Saccharomyces cerevisiae
Type F-300, lyophilized powder, activity: 130-250 units/mg protein (biuret)
H8629 Hexokinase and Glucose-6-phosphate Dehydrogenase baker's yeast (S. cerevisiae)
Lyophilized powder, hexokinase activity: ≥65 units/mg protein, glucose-6-phosphate dehydrogenase activity: ≥75 units/mg protein (b
H9408 beta;-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei
Lyophilized powder, activity: 200-750 units/mg protein
H1506 3alpha-Hydroxysteroid Dehydrogenase Pseudomonas testosteroni
Activity: ≥15 units/mg protein
I1149 Iodoacetamide
SigmaUltra
I6125 Iodoacetamide
Crystalline
K2638 Kallikrein human plasma
Buffered aqueous solution, activity: ~15 units/mg protein
K1632 Kininogen high molecular weight, Two Chain human plasma
Lyophilized powder
P4524 beta;-Lactamase from Enterobacter cloacae
Type IV, lyophilized powder, activity: 0.2-0.6 units/mg protein (using benzylpenicillin)
P4399 beta;-Lactamase from Enterobacter cloacae
Type III, lyophilized powder, activity: 6-18 units/mg protein (using benzylpenicillin)
L2395 D-Lactic Dehydrogenase Leuconostoc mesenteroides
Ammonium sulfate suspension, activity: 1,000-3,000 units/mg protein (biuret)
L1378 L-Lactic Dehydrogenase
Type X, ammonium sulfate suspension, activity: ≥600 units/mg protein
L1254 L-Lactic Dehydrogenase
Type XI, salt-free, lyophilized powder, activity: 700-1,200 units/mg protein
L2884 Leupeptin
Microbial, ≥90% (HPLC)
L8511 Leupeptin
Synthetic, ≥85% (HPLC)
L9783 Leupeptin
Microbial, ≥90% (HPLC)
L2023 Leupeptin
≥90% (HPLC), microbial
L9780 Lipase
Buffered aqueous solution, activity: ≥250 units/mg protein (Lowry)
L8906 Lipase
Lyophilized powder, activity: ≥300 units/mg solid (using olive oil)
L0382 Lipase
Type VI-S, lyophilized powder
L9518 Lipase
Type XIII, lyophilized powder, activity: ≥15 units/mg solid
L3001 Lipase
Type I, lyophilized powder, activity: ~10 units/mg protein (using triacetin, inactive on olive oil)
L7386 Lysostaphin from Staphylococcus staphylolyticus
Lyophilized powder, Protein: ~60%, activity: ≥500 units/mg protein
L4402 Lysostaphin from Staphylococcus staphylolyticus
Lyophilized powder, >97% (SDS-PAGE), Protein: ~50%, activity: ≥2,000 units/mg protein
M6159 alpha;2-Macroglobulin from human plasma
Lyophilized powder, ≥98% (SDS-PAGE)
M2634 Malic Dehydrogenase porcine heart
Buffered aqueous glycerol solution, activity: ~700 units/mg protein (biuret)
M5257 Malic Dehydrogenase (oxalacetate-decarboxylating) chicken liver
Ammonium sulfate suspension, activity: 10-30 units/mg protein (modified Warburg-Christian)
A6770 Methotrexate hydrate
M9659 4-Methylumbelliferyl N-acetyl-beta;-D-galactosaminide
≥98% (TLC)
M6018 4-Methylumbelliferyl beta;-D-cellobioside
M5510 4-Methylumbelliferyl beta;-D-fucoside
M1633 4-Methylumbelliferyl beta-D-galactopyranoside
≥99% (TLC)
M3657 4-Methylumbelliferyl alpha-D-mannopyranoside
~98% (TLC)
M0905 4-Methylumbelliferyl beta;-D-mannopyranoside
≥98% (TLC)
M8168 4-Methylumbelliferyl phosphate disodium salt
M7008 4-Methylumbelliferyl-beta-D-xylopyranoside
M9901 Mutanolysin from Streptomyces globisporus ATCC 21553
P0389 Penicillinase from Bacillus cereus
Lyophilized powder, activity: 1,500-3,000 units/mg protein (using benzylpenicillin)
P8375 Peroxidase horseradish
Type VI, essentially salt-free, lyophilized powder, activity: 250-330 units/mg solid (using pyrogallol)
P6782 Peroxidase horseradish
Type VI-A, essentially salt-free, lyophilized powder, activity: ~1000 units/mg solid (using ABTS), activity: 250-330 units/mg solid (using pyrogallol)
P8250 Peroxidase horseradish
Type II, essentially salt-free, lyophilized powder, activity: 150-250 units/mg solid (using pyrogallol)
P8125 Peroxidase horseradish
Type I, essentially salt-free, lyophilized powder, activity: 50-150 units/mg solid (using pyrogallol)
P8415 Peroxidase horseradish
Type XII, essentially salt-free, lyophilized powder, activity: 250-330 units/mg solid (using pyrogallol)
P6140 Peroxidase horseradish
Type X, ammonium sulfate suspension, activity: ~250 units/mg protein (biuret, using pyrogallol)
P1709 Peroxidase, Maleimide Activated from horseradish
Lyophilized powder, activity: >200 units/mg protein
P1146 Phosphatase, Acid
Lyophilized powder, activity: 3-10 units/mg solid
P1435 Phosphatase, Acid
Ammonium sulfate suspension, activity: ≥10 units/mg protein (modified Warburg-Christian)
P3147 Phosphatase, Acid, Prostatic
Lyophilized powder, activity: ~2 units/g solid
P1649 Phosphatase, Acid, Prostatic
Lyophilized powder, activity: ≥200 units/mg protein
P6772 Phosphatase, Alkaline
Lyophilized powder, activity: 2,000-4,000 DEA units/mg protein
P5931 Phosphatase, Alkaline
Lyophilized powder, activity: 30-60 units/mg protein (in glycine buffer)
P7634 3-Phosphoglyceric Phosphokinase from baker's yeast (S. cerevisiae)
Ammonium sulfate suspension, activity: 1,500-2,500 units/mg protein
R7385 Phosphoramidon disodium salt
≥97% (TLC)
P1867 Plasmin from human plasma
Lyophilized powder, activity: ≥3 units/mg protein
P5661 Plasminogen from human plasma
Lyophilized powder, activity: ≥2 units/mg protein, suitable for streptokinase determination by clot formation procedure
P2308 Proteinase "K" Tritirachium album
R5125 Ribonuclease A from bovine pancreas
Type III-A, ≥85% (SDS-PAGE), activity: 85-140 Kunitz units/mg protein
L9756 (−)-Tetramisole hydrochloride
≥99% (GC)
T6634 Thrombin
T3399 Thrombin
T1063 Thrombin
T2807 Thymidine Phosphorylase Escherichia coli
Recombinant, expressed in, buffered aqueous solution, ≥500 units/mL
T5398 Transglutaminase from guinea pig liver
Lyophilized powder, activity: ≥1.5 units/mg protein
T6424 Trypsin
T7409 Trypsin
T3568 TrypZean™
T3449 TrypZean™ Solution, 1+
Recombinant, expressed in, sterile-filtered
U1500 Urease Canavalia ensiformis (Jack bean)
back to top
Nucleosides, Nucleotides, and Cofactors
Catalog # Name
A8626 Adenine
≥99%
A9126 Adenine
≥99%
A8751 Adenine
≥99%
A9376 Adenosine 2′:3′-cyclic monophosphate sodium salt
~98%
A2754 Adenosine 5′-diphosphate sodium salt
Bacterial, 95-99%
A9396 Adenosine 2′-monophosphate
From yeast
A1752 Adenosine 5′-monophosphate sodium salt
From yeast, ≥99%
A8937 Adenosine 5′-triphosphate
From bacterial, Minimum 95% by HPLC
A7699 Adenosine 5′-triphosphate
SigmaUltra, ≥99%
A9062 Adenosine 5′-triphosphate
≥95%
A9187 Adenosine 5′-triphosphate
95-98%, from bacterial
FLAAM Adenosine 5′-triphosphate (ATP) assay mix
Lyophilized powder
A9384 S-(5′-Adenosyl)-L-homocysteine
Crystalline
C8631 Cytidine 3′:5′-cyclic mono­phos­phate
~98%
C7137 Cytidine 2′-monophosphate
~98%
C1133 Cytidine 3′-monophosphate
~98%
D4635 2′-Deoxycytidine 5′-triphosphate disodium salt
~97%
D5287 2′-Deoxyinosine
≥98%
D5782 2′,3′-Dideoxycytidine
≥98% (HPLC)
F6625 Flavin adenine dinucleotide disodium salt hydrate
≥95% (HPLC), powder
G9284 Guanidine hydrochloride solution
8 M in H2O (filtered)
G4505 Guanidine hydrochloride
≥99% (Cl)
G7627 Guanosine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt
~95%
H9377 Hypoxanthine
≥99%
I3500 Indoxyl acetate
≥95% (TLC)
I4125 Inosine
≥99%
I4625 Inosine 5′-monophosphate disodium salt
From yeast, Grade III, 98-100%
K3000 Kinetin riboside
M0132 5-Methyltetrahydrofolic acid disodium salt
≥88% (UV-Vis)
M6885 1-Methyluric acid
N1161 beta-NADH, disodium, hydrate, 100mg
~98%
N7004 β-Nicotinamide adenine dinucleotide
N6522 β-Nicotinamide adenine dinucleotide
N0632 β-Nicotinamide adenine dinucleotide
~95%
N0505 beta-Nicotinamide adenine dinucleotide phosphate sodium salt
~98%
N3886 beta-Nicotinamide adenine dinucleotide phosphate sodium salt
≥95%, powder
N1630 beta-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt
~95%
N7505 beta-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt
~98%
N8129 beta-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate
~98%
P3647 Phytol
P9389 Poly(deoxyguanylic-deoxycytidylic) acid sodium salt
P9755 Pyridoxine hydrochloride
F6750 Riboflavin 5′-monophosphate sodium salt dihydrate
73-79% (fluorimetric)
T3512 Thymidine 3′-monophosphate sodium salt
99% (HPLC)
U4625 Uridine 5′-diphosphoglucose disodium salt from Saccharomyces cerevisiae
≥98%
U6751 Uridine 5′-diphosphoglucuronic acid trisodium salt
98-100%
U9376 Uridine 2′-monophosphate lithium salt
~98%
U6375 Uridine 5′-monophosphate disodium salt
From yeast, ≥98%
X0626 Xanthine
≥99.5% (HPLC), purified by recrystallization
back to top
Proteins and Derivatives
Catalog # Name
A2153 Albumin
PH ~7, 1 % (w/v) aqueous solution, ≥96% (agarose gel electrophoresis), lyophilized powder
A7906 Albumin
A7030 Albumin
PH ~7, 1% 0.15 M NaCl, suitable for diluent in ELISA applications, ≥98% (agarose gel electrophoresis), lyophilized powder
A4503 Albumin
≥96% (agarose gel electrophoresis), lyophilized powder
A7888 Albumin
PH ~5.2, 1% aqueous solution, ≥96% (agarose gel electrophoresis), lyophilized powder
A9647 Albumin
Heat-shocked fractionate, pH ~7, 1 % (w/v) aqueous solution, ≥96% (agarose gel electrophoresis), lyophilized powder
A3294 Albumin
Further purified Fraction V, ≥98% (agarose gel electrophoresis), lyophilized powder
A6003 Albumin
Essentially fatty acid free, ≥96% (agarose gel electrophoresis), lyophilized powder
A3059 Albumin
Further purified Fraction V, ~99% (agarose gel electrophoresis), lyophilized powder, Essentially γ-globulin free
A7638 Albumin
Further purified, ≥99% (agarose gel electrophoresis), lyophilized powder
A0281 Albumin
Essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis), lyophilized powder
A8022 Albumin
PH ~5.4, 1 % (w/v) aqueous solution, ≥96% (agarose gel electrophoresis), lyophilized powder
A3912 Albumin
PH ~5.2, 1 % (w/v) aqueous solution, ≥96% (agarose gel electrophoresis), lyophilized powder
A3803 Albumin
Further purified, pH ~7, 1 % (w/v) aqueous solution, suitable for diluent in ELISA applications, ≥98% (agarose gel electrophoresis), lyophilized powder
A4378 Albumin
1× Crystallized, ≥97% (agarose gel electrophoresis), lyophilized powder
A7511 Albumin
Essentially fatty acid free, ≥97% (agarose gel electrophoresis), lyophilized powder
B6917 Albumin
For molecular biology, Non-acetylated, lyophilized powder
A3902 Albumin
A7284 Albumin
A7409 Albumin
A7034 Albumin
A8327 Albumin
A1662 Albumin
A9263 Albumin
A5503 Albumin
Grade V, ≥98% (agarose gel electrophoresis), lyophilized powder
A2512 Albumin
Grade VI, ≥98% (agarose gel electrophoresis), lyophilized powder
A1653 Albumin
96-99% (agarose gel electrophoresis)
A6909 Albumin
30% in 0.85% sodium chloride
A6784 Albumin
10% in 0.85% sodium chloride and 0.05% sodium azide
A4414 Albumin
~99% (agarose gel electrophoresis), lyophilized powder
A1830 Albumin
≥98% (agarose gel electrophoresis)
B8894 Albumin, Acetylated from bovine serum
20 mg/mL in H2O, for molecular biology
C5890 Casein
Suitable for substrate for protein kinase (after dephosphorylation), purified powder
C3400 Casein
Powder
C8654 Casein
C4765 Casein solution from bovine milk
5%
C0755 Conalbumin chicken egg white
Substantially iron-free
C2037 Cytochrome c
≥95% as based on Mol. Wt. 12,327
C7752 Cytochrome c
≥95% as based on Mol. Wt. 12,384
C2506 Cytochrome c
≥95% (SDS-PAGE)
C2867 Cytochrome c
≥99% (SDS-PAGE)
F8630 Fibrinogen
Type I-S, ~75% protein (≥75% of protein is clottable)
F4753 Fibrinogen
~60% protein (~ 95% of protein is clottable)
F3879 Fibrinogen
~60% protein (>80% of protein is clottable)
F4883 Fibrinogen
35-65% protein (~95% of protein is clottable).
G7765 Gelatin
G6144 Gelatin
G3375 Gliadin from wheat
G5009 γ-Globulins
99% (agarose gel electrophoresis)
G7516 γ-Globulins
NaCl ≤5%
G9513 γ-Globulins
99% (agarose gel electrophoresis)
G4386 γ-Globulins
99% (electrophoresis)
G2018 γ-Globulins
~99% (agarose gel electrophoresis)
G5640 Globulins goat
Cohn fraction II, III
G2388 Globulins Cohn fraction II, III human
Lyophilized powder
G3387 Globulins Cohn fraction IV human
H8283 Hemocyanin
PBS solution
B8556 Hemocyanin
Buffered aqueous solution
K0253 Keratin solution from human epidermis
Urea solution
L6385 alpha;-Lactalbumin from bovine milk
For use as a marker in SDS-PAGE
L5137 beta;-Lactoglobulin A from bovine milk
Isoelectric focusing marker, pI 5.1
M1778 Mucin from porcine stomach
Type III, partially purified powder
M2941 Myelin Basic Protein mouse
>95% (SDS-PAGE), >95% (confirmed by immuno-blotting), lyophilized powder (salt free)
M1882 Myoglobin
≥90% (PhastGel electrophoresis), essentially salt-free, lyophilized powder
M0630 Myoglobin
95-100%, essentially salt-free, lyophilized powder
M6036 Myoglobin
≥95% (SDS-PAGE), buffered aqueous glycerol solution
M7527 Myoglobin
Recombinant, expressed in, 95-100%, buffered aqueous solution
P7000 Pepsin porcine gastric mucosa
P6887 Pepsin porcine gastric mucosa
P7012 Pepsin porcine gastric mucosa
P7125 Pepsin porcine gastric mucosa
P7750 Peptone from animal tissue
From meat, Type I
P1286 B-Phycoerythrin from Porphyridium cruentum
Lyophilized powder
P5115 Polypep® Low Viscosity
P1742 Prealbumin from human plasma
C8754 Thiamine pyrophosphate
≥97%
T2275 Troponin from porcine muscle
T9924 Troponin I from human heart
Lyophilized powder
T0175 Troponin T from human heart
≥60% (SDS-PAGE), lyophilized powder
back to top
Substrates
Catalog # Name
A2056 Acetyl coenzyme A
~95%, powder
A2181 Acetyl coenzyme A
~95% (HPLC)
A9891 7-Amino-4-methylcoumarin
A3261 Arg-Arg beta-naphthylamide trihydrochloride
A9941 2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt
Tablet, 10 mg substrate per tablet
A2765 Azocasein
B1911 BCIP/NBT Liquid Substrate System
B4750 Nalpha-Benzoyl-DL-arginine beta-naphthylamide hydrochloride
B1260 N-Benzoyl-Phe-Val-Arg 4-methoxy-beta;-naphthylamide hydrochloride
B7632 N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride
N3002 Bis(p-nitrophenyl) phosphate sodium salt
O3629 Bis(2,4,6-trichlorophenyl) oxalate
G1013 Gly-Arg 4-methoxy-beta;-naphthylamide dihydrochloride
H9882 4-Hydroxybenzhydrazide
≥97%
K3752 α-Ketoglutaric acid
~95%
K2000 α-Ketoglutaric acid
≥98% (enzymatic)
K1875 α-Ketoglutaric acid
≥98% (enzymatic)
K2010 α-Ketoglutaric acid
SigmaUltra
L0376 L-Leucine beta-naphthylamide hydrochloride
L2158 L-Leucine p-nitroanilide hydrochloride
M3167 4-Methylumbelliferyl alpha;-L-arabinopyranoside
N9003 4-Nitrophenyl N-acetyl-beta;-D-galactosaminide
≥98%
N9376 4-Nitrophenyl N-acetyl-beta;-D-glucosaminide
98-100%
N3512 4-Nitrophenyl alpha;-L-arabinopyranoside
N5759 4-Nitrophenyl beta;-D-cellobioside
N3628 4-Nitrophenyl alpha-L-fuco­pyran­oside
≥98% (TLC)
N3378 4-Nitrophenyl beta-D-fuco­pyran­oside
≥98% (TLC)
N3016 p-Nitrophenyl galacto-N-bioside
≥98%
N0877 4-Nitrophenyl alpha-D-galacto­pyran­oside
N1252 4-Nitrophenyl beta-D-galacto­pyran­oside
≥98% (enzymatic)
N8016 2-Nitrophenyl beta-D-glucopyranoside
Powder
N1377 4-Nitrophenyl alpha;-D-glucopyranoside
≥99%
N7006 4-Nitrophenyl beta;-D-glucopyranoside
98% by TLC
N1752 4-Nitrophenyl beta;-D-lactopyranoside
Powder
N2127 4-Nitrophenyl alpha-D-mannopyranoside
N3129 4-Nitrophenyl phosphate
N9389 4-Nitrophenyl phosphate
Tablet
N2765 4-Nitrophenyl phosphate
Tablet
N2640 4-Nitrophenyl phosphate
Tablet
N2132 4-Nitrophenyl beta-D-xylopyranoside
≥98%
P7288 o-Phenylenediamine dihydrochloride
Tablet, 20 mg substrate per tablet
P8412 o-Phenylenediamine dihydrochloride
Tablet, 30 mg substrate per tablet
P1526 o-Phenylenediamine dihydrochloride
Crystalline
R2886 Resorufin beta;-D-glucuronide sodium salt
≥90%
N1891 SIGMAFAST™ p-Nitrophenyl phosphate Tablets
Tablet, To prepare 5 mL
P9187 SIGMAFAST™ OPD
Tablet
860336 3,3′,5,5′-Tetramethylbenzidine
T2885 3,3′,5,5′-Tetramethylbenzidine
≥98% (TLC)
87748 3,3′,5,5′-Tetramethylbenzidine
T8768 3,3′,5,5′-Tetramethylbenzidine
Bulk package
87750 3,3′,5,5′-Tetramethylbenzidine
T8665 3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System
back to top