Extranet 
Welcome to SAFC Supply Solutions
 New Proligo® Reagents
SAFC Supply Solutions®
New Products:
 2’Fluoro Phosphoramidites  6-FAM
2’Fluoro Phosphoramidites
Download Literature: 2'Fluoro Phosphoramidites (425 Kb PDF)
As a complement to our existing amidite products, SAFC Supply Solutions adds 2’Fluoro A and G monomers.
DMT-2'Fluoro Phosphoramidites structures image
Key Features of 2’Fluoro Amidites
2'Fluoro amidites are used to synthesize oligonucleotides that are more thermally stable and provide increased nuclease resistance
All four bases are available:
• DMT-2'Fluoro-dA(bz) Phosphoramidite
• DMT-2'Fluoro-dC(ac) Phosphoramidite
• DMT-2'Fluoro-dG(ib) Phosphoramidite
• DMT-2'Fluoro-dT Phosphoramidite
2'FluoroAmidites can be employed together with DNA and RNA amidites
Compatible with deprotection schemes based on methylamine or AMA (conc. aqueous ammonia/40% aqueous methylamine 1/1, v/v)
Lot to lot consistent purity and performance
Manufactured under a certified ISO 9000:2000 quality system
Analytical Specifications of 2'Fluoro Amidites
Test 2'F-dA(bz) 2'F-dC(ac) 2'F-dG(ib) 2'F-dU
Reverse Phase HPLC ≥98.0% ≥98.0% ≥98.0% ≥98.0%
31P-NMR ≥98% ≥99% ≥98% ≥99%
Ordering Information
 DMT-2'Fluoro Phosphoramidites
 Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
A211281-01 DMT-2'Fluoro-dA(bz) Amidite 1 x 0.25g
C213281-01 DMT-2'Fluoro-dC(ac) Amidite 1 x 0.25g
G211281-01 DMT-2'Fluoro-dG(ib) Amidite 1 x 0.25g
U211281-01 DMT-2'Fluoro-dU Amidite 1 x 0.25g
 Compatible with ABI® Instruments
A211231-01 DMT-2'Fluoro-dA(bz) Amidite 1 x 0.25g
C213231-01 DMT-2'Fluoro-dC(ac) Amidite 1 x 0.25g
G211231-01 DMT-2'Fluoro-dG(ib) Amidite 1 x 0.25g
U211231-01 DMT-2'Fluoro-dU Amidite 1 x 0.25g
Please contact your local sales office for availability and packaging in your country.
6-FAM
Download Literature: 6-FAM (414 Kb PDF)
6-FAM as a common fluorescent derivatization reagent for covalent labelling of synthetic oligonucleotides.
6-FAM structure image
Key Features of 6-FAM
6-FAM consists of one isomer derived from 6-carboxy fluorescein
Couples to the 5'-end of the oligonucleotide using standard synthesis protocols
Protecting groups on the fluorescein moiety are removed under standard conditions of cleavage and deprotection with concentrated ammonia
A detritylation step is not required since the fluorescein phosphoramidite does not contain a DMT-group
The labeled oligonucleotide is ready for use in most applications after the evaporation of ammonia—standard procedures can be used if additional purification is required
Lot-to-lot consistent purity and performance
Manufactured under a certified ISO 9000:2000 quality system
Analytical Specifications of 6-FAM
Test 6-FAM
HPLC Purity ≥97.0%
31P-NMR ≥97.0%
Ordering Information
 6-FAM
 Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
X010181-01 6-FAM 1 x 0.1g
 Compatible with ABI® Instruments
X010131-01 6-FAM 1 x 0.1g
Please contact your local sales office for availability and packaging in you country.