|
Biotin Phosphoramidite
| Product Breadth of Proligo Reagents |
|
| Product Breadth of Proligo Reagents |
|
| Biotin-labeled oligonucleotides have become extremely popular in DNA capture and detection applications, due to their very high sensitivity and strong, specific binding affinity with avidin and streptavidin proteins. Biotin-based detection methods are widely used by researchers. Biotin labeling is compatible with PCR and most hybridization techniques. |
| Key Features of the Biotin Phosphoramidite |
|
|
Behaves like any standard amidite on the DNA synthesizer |
|
|
Readily soluble in acetonitrile |
|
|
Biotin compound has a dimethozytrityl (DMT) — group on the ring structure, which allows coupling-yield determination and trityl-selective purification |
|
|
A long, water-compatible linker arm ensures maximum sensitivity |
|
|
The labeled oligonucleotide is ready for use in most applications after the evaporation of ammonia. Standard procedures can be used if additional purification is required |
|
|
Provides a coupling efficiency of 95% |
|
|
Download Literature: User Instructions Biotin Phosphoramidite (670 Kb PDF)
View Structure: DMT-Biotin-β-Cyanoethy-lphosphoramidite |
| Labels and Modifications - Biotin Phosphoramidite |
| Compatible with Expedite™ and Polygen® Instruments |
| Product No. |
Description |
Unit |
| M010381-01 |
Biotin amidite |
0.1g |
| M010382-01 |
Biotin amidite |
0.25 g |
| Compatible with ABI™ Instruments |
| M010331-01 |
Biotin amidite |
0.1g |
| M010332-01 |
Biotin amidite |
0.25 g |
| Custom Products |
Phosphoramidites of non-catalogue linkers, reporters or modifiers are offered as custom synthesis products. Please enquire. |
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP. © 2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission. Other trademarks are the property of their respective owners.