Extranet 
Welcome to SAFC Supply Solutions
Controlled Pore Glass (Free flow CPG)
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites:
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
2'Fluoro Phosphoramidites
DMT-2'Fluoro Phosphoramidites
5'-NPPOC Phosphoramidites
 Fast Deprotection Chemistries:
DNA
RNA
 Supports:
Controlled Pore Glass (CPG)
Columns
 Solvents:
Liquid Reagents
Activator 42™
 Modifications:
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
6-FAM
 Linkers:
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
Since Merrifield first described solid phase synthesis, researchers have investigated numerous supports for the chemical synthesis of DNA. Controlled Pore Glass (CPG) has proven to be the most accepted standard, with features such as high surface area, efficient mass transfer and chemical inertness.
Because the pore density of CPG can be adjusted, the solid support can be tailored to the size of the synthetic oligonucleotide required. Bigger pore size is typically preferable in minimizing steric effects during the synthesis of long oligonucleotides, while smaller pore size fosters improved synthesis consistency.
Our process guarantees consistent production of CPGs to specifications designed for DNA synthesis.
All products are tested in our Quality Control labs for purity and identity prior to release. SAFC Proligo Reagent’s facility in Hamburg, Germany is ISO 9001 certified.
 On this Page
DNA CPG
Fast deprotection DNA CPG
RNA CPG
Amino-ON CPG
Universal CPG
DNA CPG
Analytical Parameters of CPG
  CPG (UHL) CPG 500Å CPG 1000Å
Pore size (nm) 33 50 100
Pore volume (ml/g) 1.3 1.3 1.3
Grain size (µm) 100 - 180 100 – 180 100 - 180
Surface (m2/g) ~100 ~60 ~30
Nucleoside-Loaded CPG
  Loading µmol/g
CPG 500Å 30-40
CPG 1000Å 25-35
CPG UHL 50-70
CPG UHL 90-110
DNA Controlled Pore Glass
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
  500 Å, extent of labeling: 30-40 μmol/g
A301001-01 dA(bz)-CPG 1 g
A301010-01 dA(bz)-CPG 10 g
  1000 Å, extent of labeling: 25-35 μmol/g
A401001-01 dA(bz)-CPG 1 g
A401010-01 dA(bz)-CPG 10 g
  500 Å, extent of labeling: 30-40 μmol/g
C301001-01 dC(bz)-CPG 1 g
C301010-01 dC(bz)-CPG 10 g
  1000 Å, extent of labeling: 25-35 μmol/g
C401001-01 dC(bz)-CPG 1 g
C401010-01 dC(bz)-CPG 10 g
  500 Å, extent of labeling: 30-40 μmol/g
G301001-01 dG(ib)-CPG 1 g
G301010-01 dG(ib)-CPG 10 g
  1000 Å, extent of labeling: 25-35 μmol/g
G401001-01 dG(ib)-CPG 1 g
G401010-01 dG(ib)-CPG 10 g
  500 Å, extent of labeling: 30-40 μmol/g
T301001-01 dT-CPG 1 g
T301010-01 dT-CPG 10 g
  1000 Å, extent of labeling: 25-35 μmol/gg
T401001-01 dT-CPG 1 g
T401010-01 dT-CPG 10 g
Custom Products Bulk packaging up to multiple kg per container and customized packaging with alternative quantities of amidites are available upon request.
  CPG-products can be provided with higher loadings based on UHL-CPG (50-70 µmol/g or 90-110 µmol/g), please enquire.
Fast deprotection DNA CPG
Fast Deprotection CPG
Product No. Description Unit
A302001-01 CPG 500Å, dA(tac), 30-40 μmol/g 1 x 1g
C302001-01 CPG 500Å, dC(tac), 30-40 μmol/g 1 x 1g
G302001-01 CPG 500Å, dG(tac), 30-40 μmol/g 1 x 1g
A402001-01 CPG 1000Å, dA(tac), 25-35 μmol/g 1 x 1g
C402001-01 CPG 1000Å, dC(tac), 25-35 μmol/g 1 x 1g
G402001-01 CPG 1000Å, dG(tac), 25-35 μmol/g 1 x 1g
C302010-01 CPG 500Å, dC(tac), 30-40 μmol/g 1 x 10g
C402010-01 CPG 500Å, dC(tac), 25-35 μmol/g 1 x 10g
G305001-01 CPG 500Å, dG(tac), 30-40 μmol/g 1 x 1g
G305010-01 CPG 500Å, dG(tac), 30-40 μmol/g 1 x 10g
RNA CPG
Fast Deprotection RNA CPG
Product No. Description Unit
A602001-01 CPG 500Å, rA(tac), 25-35 μmol/g 1 x 1g
C602001-01 CPG 500Å, rC(tac), 25-35 μmol/g 1 x 1g
G602001-01 CPG 500Å, rG(tac), 25-35 μmol/g 1 x 1g
U601001-01 CPG 500Å, rU, 25-35 μmol/g 1 x 1g
2'0-Methyl RNA CPG
Product No. Description Unit
A601101-01 CPG 500Å 2'O-Methyl-rA(bz), 25-35 μmol/g 1 x 1g
C602101-01 CPG 500Å 2'O-Methyl-rC(tac), 25-35 μmol/g 1 x 1g
G601101-01 CPG 500Å 2'O-Methyl-rG(ib), 25-35 μmol/g 1 x 1g
U601101-01 CPG 500Å 2;O-Methyl-rU, 25-35 μmol/g 1 x 1g
Amino-ON CPG
Amino-ON CPG can be employed to conjugate biotin, fluorescein or other modifiers and reporter groups to the 3'-end of oligonucleotides or to attach oligonucleotides to surfaces. Amino-ON CPG from SAFC Proligo Reagents combines the advanced features of cleavage and deprotection under mild conditions, and maintenance of the integrity of the 3'-modification with high synthesis yields.  
  Download Literature:
  •  User Instructions
Amino-ON CPG
(667k pdf)
  View Structure:
  •  Amino-ON CPG
Key Features of 3’-Amine Oligonucleotides
Selective conjugation to reporter groups, purification handles or other modifiers
3'-end attachment to electrophilic surfaces
Offers protection against exonucleolytic degradation
These features enable 3’-amine oligonucleotides to be used in applications such as 3’-labeling with base sensitive dyes; 3’-biotin modifications; dual-labeled probes (eg TaqMan® probes and Molecular beacons); and microarray spotting.  
Key Features of Amino-ON CPG
Introduces a primary amino group attached to a C6-linker
Fully compatible with standard phosphoramidite reagents and synthesis conditions
Can be employed together with DNA or RNA phosphoramidites in the same synthesis to produce mixmer oligonucleotides
Can be used with DNA, RNA and Locked Nucleic Acid® (LNA®); 5'-oligonucleotide modifications such as fluorescein or biotin; and with the synthesis of thioated DNA oligonucleotides
Compatible with dG(dmf) fast deprotection chemistry: the oligonucleotide can be cleaved from the support and deprotected in concentrated ammonia at 55°C in 2 hours
Provides all the advantages of homogeneous pore size porous glass supports
Standard pore size of 500Å
Loading of 30-40 µmol/g
Coupling performance of ≥ 99.0% stepwise efficiency
DMT-protected in the same way as standard nucleoside-loaded CPG
Amino on CPG
  500 Å, extent of labeling: 30‑40 μmol/g
M020101-01 Amino-ON CPG  
Universal CPG
Universal CPG from SAFC Proligo Reagents is truly universal as it can be employed in the synthesis of any oligonucleotide sequence instead of conventional nucleoside loaded CPG - whether loaded with adenosine, cytidine, guanosine or thymidine nucleosides. The use of universal CPG releases your resources as the purchase, handling and storage of differentially loaded types of nucleoside-CPG is no longer required. And 3’-end synthesis errors, because of accidental interchange of polymeric supports, are essentially eliminated.  
  Download Literature:
  •  User Instructions
Universal CPG
(659k pdf)
  View Structure:
  •  Universal CPG
Key Features of Universal CPG
Fully compatible with standard phosphoramidite reagents and synthesis conditions
Can be employed in the synthesis of many different types of oligonucleotides:
DNA oligonucleotides
Phosphorothioates
2’O-Methyl oligonucleotides
Locked Nucleic Acid® (LNA®) oligonucleotides
Fluorescein, tetrachloro-fluorescein and biotin modifications
Particularly useful for high-throughput oligonucleotide synthesis
Coupling efficiency of ≥ 99.0% is routinely obtained
Convenient cleavage and deprotection
Avoids the use of unusual and potentially harmful reagents in the deprotection process e.g. heavy metals, sulfides
Universal CPG
Product No. Description Unit
  500 Å, extent of labeling: 30-40 μmol/g
M301001-01 Universal CPG 1 g
M301010-01 Universal CPG 10 g
  1000 Å, extent of labeling: 25-35 μmol/g
M401001-01 Universal CPG 1 g
M401010-01 Universal CPG 10 g
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP.
©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.
To order or speak with a Customer Service Representative, contact your local SAFC office.