|
DNA Phosphoramidites
| Product Breadth of Proligo Reagents |
|
| Product Breadth of Proligo Reagents |
|
Highest Quality
| SAFC Proligo Reagents’ world-class expertise in nucleic acid synthesis began in 1983 when Dr. Hubert Köster first commercialized the synthesis of ß-cyanoethylphosphoramidites. Since then, the company has refined and perfected the production of DNA and RNA phosphoramidites. Today, SAFC Proligo Reagents® offers the highest quality phosphoramidites in the industry. |
|
|
Key Features of DNA Phosphoramidites
|
|
Exocyclic amine functions are protected by a benzoyl group (dA(bz) and dC(bz)) or isobutyryl group (dG(ib)) |
|
Recommended cleavage and deprotection conditions are 8 hours at 55°C or 24 hours at room temperature using concentrated ammonia solution, for standard baseprotected oligonucleotides |
|
The high coupling efficiency of Proligo Reagents’ DNA phosphoramiditesleads to high-yield, high-quality oligonucleotides |
|
|
|
| Standard DNA Phosphoramidites |
| Compatible with Expedite™ and Polygen® Instruments |
| Product No. |
Description |
Unit |
| A111081-12 |
DMT-dA(bz) amidite |
12 × 1 g |
| A111082-12 |
DMT-dA(bz) amidite |
12 × 2 g |
| C111081-12 |
DMT-dC(bz) amidite |
12 × 1 g |
| C111082-12 |
DMT-dC(bz) amidite |
12 × 2 g |
| G111081-12 |
DMT-dG(ib) amidite |
12 × 1 g |
| G111082-12 |
DMT-dG(ib) amidite |
12 × 2 g |
| T111081-12 |
DMT-dT amidite |
12 × 1 g |
| T111082-12 |
DMT-dT amidite |
12 × 2 g |
| Standard DNA Phosphoramidites |
| Compatible with ABI™ Instruments |
| Product No. |
Description |
Unit |
| A111031-12 |
DMT-dA(bz) amidite |
12 × 1 g |
| C111031-12 |
DMT-dC(bz) amidite |
12 × 1 g |
| G111031-12 |
DMT-dG(ib) amidite |
12 × 1 g |
| T111031-12 |
DMT-dT amidite |
12 × 1 g |
| A111032-12 |
DMT-dA(bz) amidite |
12 × 2 g |
| C111032-12 |
DMT-dC(bz) amidite |
12 × 2 g |
| G111032-12 |
DMT-dG(ib) amidite |
12 × 2 g |
| T111032-12 |
DMT-dT amidite |
12 × 2 g |
| A111064-12 |
DMT-dA(bz) amidite |
12 × 4 g |
| C111064-12 |
DMT-dC(bz) amidite |
12 × 4 g |
| G111064-12 |
DMT-dG(ib) amidite |
12 × 4 g |
| T111064-12 |
DMT-dT amidite |
12 × 4 g |
| Compatible with Mermade™ Instruments (8oz 28/400 bottle) |
| Product No. |
Description |
Unit |
| A111085-06 |
DMT-dA(bz) amidite |
6 × 5 g |
| C111085-06 |
DMT-dC(bz) amidite |
6 × 5 g |
| G111085-06 |
DMT-dG(ib) amidite |
6 × 5 g |
| T111085-06 |
DMT-dT amidite |
6 × 5 g |
| A111028-06 |
DMT-dA(bz) amidite |
6 × 10 g |
| C111028-06 |
DMT-dC(bz) amidite |
6 × 10 g |
| G111028-06 |
DMT-dG(ib) amidite |
6 × 10 g |
| T111028-06 |
DMT-dT amidite |
6 × 10 g |
| Compatible with AKTA™ Oligo Pilot Instruments (100ml septum bottle) |
| Product No. |
Description |
Unit |
| A111005-01 |
DMT-dA(bz) amidite |
5 g |
| A111005-06 |
DMT-dA(bz) amidite |
6 × 5 g |
| A111010-01 |
DMT-dA(bz) amidite |
10 g |
| A111020-06 |
DMT-dA(bz) amidite |
6 × 20 g |
| C111005-01 |
DMT-dC(bz) amidite |
5 g |
| C111005-06 |
DMT-dC(bz) amidite |
6 × 5 g |
| C111010-01 |
DMT-dC(bz) amidite |
10 g |
| C111020-06 |
DMT-dC(bz) amidite |
6 × 20 g |
| G111005-01 |
DMT-dG(ib) amidite |
5 g |
| G111005-06 |
DMT-dG(ib) amidite |
6 × 5 g |
| G111010-01 |
DMT-dG(ib) amidite |
10 g |
| G111020-06 |
DMT-dG(ib) amidite |
6 × 20 g |
| T111005-01 |
DMT-dT amidite |
5 g |
| T111005-06 |
DMT-dT amidite |
6 × 5 g |
| T111010-01 |
DMT-dT amidite |
10 g |
| T111020-06 |
DMT-dT amidite |
6 × 20 g |
| Bulk Quantities (16oz 28/400 bottle) |
| Product No. |
Description |
Unit |
| A111005-01 |
DMT-dA(bz) amidite |
5 g |
| A111005-06 |
DMT-dA(bz) amidite |
6 × 5 g |
| A111010-01 |
DMT-dA(bz) amidite |
10 g |
| A111020-06 |
DMT-dA(bz) amidite |
6 × 20 g |
| C111005-01 |
DMT-dC(bz) amidite |
5 g |
| C111005-06 |
DMT-dC(bz) amidite |
6 × 5 g |
| C111010-01 |
DMT-dC(bz) amidite |
10 g |
| C111020-06 |
DMT-dC(bz) amidite |
6 × 20 g |
| G111005-01 |
DMT-dG(ib) amidite |
5 g |
| G111005-06 |
DMT-dG(ib) amidite |
6 × 5 g |
| G111010-01 |
DMT-dG(ib) amidite |
10 g |
| G111020-06 |
DMT-dG(ib) amidite |
6 × 20 g |
| T111005-01 |
DMT-dT amidite |
5 g |
| T111005-06 |
DMT-dT amidite |
6 × 5 g |
| T111010-01 |
DMT-dT amidite |
10 g |
| T111020-06 |
DMT-dT amidite |
6 × 20 g |
| Custom Products |
Bulk packaging up to multiple kg per container and customized packaging with alternative quantities of amidites are available upon request. |
SAFC and SAFC Proligo Reagents are trademarks of Sigma-Aldrich Biotechnology, LP. ©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission. Other trademarks are the property of their respective owners. |