1. TAC Chemistry
|
Substitution of standard protecting groups with the labile TAC (tert.butylphenoxyacetyl) protecting group results in ultra-fast and easy deprotection under mild conditions, suitable for oligonucleotides with base-labile monomers and reporters as well as in-situ synthesis schemes on glass surfaces.
|
| Key features of TAC Chemistry. |
|
Deprotection of the TAC group is ultra-fast: complete deprotection in concentrated ammonia occurs within 15 minutes at 55°C or two hours at room temperature |
|
TAC protecting groups are compatible with the AMA deprotection reagent (a mixture of ≥ 25% ammonia in water with 40% aqueous methylamine I/I, v/v) |
|
TAC protected amidites are highly soluble in acetonitrile. There is no need to add co-solvents such as dimethylformamide or methylene chloride |
|
TAC monomers are suitable for the synthesis of oligomers with base-labile units e.g. dyes and modifiers, because of less exposure to ammonia and the possibility of room temperature deprotection |
|
No change is required in the reagents commonly used for DNA synthesis, except that Proligo Reagents’ Fast Deprotection CAP A solution is used instead of Cap A solution |
|
The application of dA(tac) minimizes depurination and improves the quality of oligonucleotides |
| |
|
2. dG(dmf) Method
|
Changing the dG protecting group to the dimethylformamidine (dmf) base-protecting group enables rapid synthesis of high-purity, high-yield oligonucleotide, thus increasing the efficiency of high-throughput production.
|
| Key Features of dG(dmf) |
|
dG(dmf) is deprotected faster than the conventional dG(ib): the deprotection time in concentrated ammonia is reduced to 2 hours at 55°C or 1 hour at 65°C |
|
The dG(dmf)-monomer is especially suitable for G-rich sequences: incomplete deprotection is greatly reduced in comparison with the conventional dG(ib)-monomer |
|
dG(dmf)-amidite is as stable in solution as the standard dA(bz)-, dC(bz)- and dT-amidites. |
|
dG(dmf)-amidite can directly substitute for dG(ib)-amidite |
|
No change is required in the reagents commonly used for DNA synthesis (except a low concentration iodine oxidizer i.e. 0.02M in iodine, should be employed) |
| |
|
3. Substituting the dC Protecting Group
|
Changing the dC protecting group to the TAC protecting group leads to rapid synthesis of high-purity and high-yield oligonucleotides. Substituting the commonly employed dC(bz) monomer by the dC(tac) monomer enables the application of ultra-fast deprotection with the AMA reagent and provides a high-throughput method of oligonucleotide synthesis.
|
| Key Features of Substituting the dC Protecting Group |
|
The deprotection of oligonucleotide synthesis products with the AMA reagent is ultra-fast: complete deprotection requires 10 minutes at 65°C |
|
Side reactions at C-monomers through transamination are eliminated |
|
Not compatible with some base-labile modified nucleosides |
|
dC(tac)-amidite can directly substitute for dC(bz)-amidite |
|
No change is required in the reagents commonly used for DNA synthesis: acetonitrile is used to dissolve the amidite. The standard acetic anhydride capping reagent can be employed |
| |
|
| FAST Deprotection Phosphoramidites |
| Compatible with Expedite™ and Polygen® Instruments |
| Product No. |
Description |
Unit |
| A112081-12 |
DMT-dA(tac) amidite |
12 × 1 g |
| A112082-12 |
DMT-dA(tac) amidite |
12 × 2 g |
| C112081-12 |
DMT-dC(tac) amidite |
12 × 1 g |
| C112082-12 |
DMT-dC(tac) amidite |
12 × 2 g |
| G115081-12 |
DMT-dG(dmf) amidite |
12 × 1 g |
| G115082-12 |
DMT-dG(dmf) amidite |
12 × 2 g |
| G112081-12 |
DMT-dG(tac) amidite |
12 × 1 g |
| G112082-12 |
DMT-dG(tac) amidite |
12 × 2 g |
| Compatible with ABI™ Instruments |
| Product No. |
Description |
Unit |
| A112031-12 |
DMT-dA(tac) amidite |
12 × 1 g |
| A112032-12 |
DMT-dA(tac) amidite |
12 × 2 g |
| C112031-12 |
DMT-dC(tac) amidite |
12 × 1 g |
| C112032-12 |
DMT-dC(tac) amidite |
12 × 2 g |
| G115031-12 |
DMT-dG(dmf) amidite |
12 × 1 g |
| G115032-12 |
DMT-dG(dmf) amidite |
12 × 2 g |
| G112031-12 |
DMT-dG(tac) amidite |
12 × 1 g |
| G112032-12 |
DMT-dG(tac) amidite |
12 × 2 g |
| Compatible with MerMade™ Instruments (8oz 28/400 bottle) |
| Product No. |
Description |
Unit |
| C112081-12 |
DMT-dC(tac) amidite |
12 × 1 g |
| C112082-12 |
DMT-dC(tac) amidite |
12 × 2 g |
| G115028-06 |
DMT-dG(dmf) amidite |
|
| Compatible with AKTA™ oligopilot Instruments (100ml septum bottle) |
| Product No. |
Description |
Unit |
| A112010-01 |
DMT-dA(tac) amidite |
|
| C112010-01 |
DMT-dC(tac) amidite |
10 g |
| C112020-06 |
DMT-dC(tac) amidite |
6 × 20 g |
| G115005-01 |
DMT-dG(dmf) amidite |
5 g |
| G112010-01 |
DMT-dG(tac) amidite |
| Bulk Quantities (16oz 28/400 bottle) |
| Product No. |
Description |
Unit |
| C112010-01 |
DMT-dC(tac) amidite |
10 g |
| C112020-06 |
DMT-dC(tac) amidite |
6 × 20 g |
| G115021-06 |
DMT-dC(tac) amidite |
6 × 10g |
| FAST Deprotection CPG |
| Product No. |
Description |
Unit |
| |
500 Å, extent of labeling: 30-40 μmol/g |
| A302001-01 |
dA(tac)-CPG |
|
| |
1000 Å, extent of labeling: 25-35 μmol/g |
| A402001-01 |
dA(tac)-CPG |
|
| |
500 Å, extent of labeling: 30-40 μmol/g |
| C302001-01 |
dC(tac)-CPG |
1 g |
| C302010-01 |
dC(tac)-CPG |
10 g |
| |
500 Å, extent of labeling: 30-40 μmol/g |
| G305001-01 |
dG(dmf)-CPG |
1 g |
| G305010-01 |
dG(dmf)-CPG |
10 g |
| |
500 Å, extent of labeling: 30-40 μmol/g |
| G302001-01 |
dG(tac)-CPG |
|
| |
1000 Å, extent of labeling: 25-35 μmol/g |
| G402001-01 |
dG(tac)-CPG |
|
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP. ©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission. Other trademarks are the property of their respective owners. |
|