Extranet 
Welcome to SAFC Supply Solutions
Fluorescein Phosphoramidite
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites:
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
2'Fluoro Phosphoramidites
DMT-2'Fluoro Phosphoramidites
5'-NPPOC Phosphoramidites
 Fast Deprotection Chemistries:
DNA
RNA
 Supports:
Controlled Pore Glass (CPG)
Columns
 Solvents:
Liquid Reagents
Activator 42™
 Modifications:
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
6-FAM
 Linkers:
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
Fluorescein phosphoramidite is composed of a protected fluorescein molecule linked to a ß-cyanoethylphosphoramidite. The fluorescein derivative consists of to isomers derived form 5- and 6- carboxy fluorescein. As a result, two peaks will be seen on a RP-HPLC chromatogram of the synthesis product.  
  Download Literature:
  •  User Instructions
Fluorescein Phosphoramidite
(656k pdf)
  View Structure:
  •  Fluorescein-ß-
Cyanoethyl
phosphoramidite
Key Features of the Fluorescein Phosphoramidite  
Readily soluble in acetonitrile  
Couples to the 5’ end of the oligonucleotide using standard synthesis protocols
Protecting groups on the fluorescein moiety are removed under standard conditions of cleavage and deprotection with concentrated ammonia
A detritylation step is not required since the fluorescein phosphoramidite does not contain a dimethoxytrityl (DMT-) group
The labeled oligonucleotide is ready for use in most applications after the evaporation of ammonia. Standard procedures can be used if additional purification is required
The fluorescein moiety provides a purification handle in RP-HPLC purification
Labels and Modifications - Fluorescein Phosphoramidite
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
M010181-01 Fluorescein amidite 1 x 0.1g
Compatible with ABI™ Instruments
M010131-01 Fluorescein amidite 1 x 0.1g
Custom Products Phosphoramidites of non-catalogue linkers, reporters or modifiers are offered as custom synthesis products. Please enquire.
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP.
©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.
To order or speak with a Customer Service Representative, contact your local SAFC office.