Extranet 
Welcome to SAFC Supply Solutions
Fast Deprotection RNA Phosphoramidites
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites:
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
2'Fluoro Phosphoramidites
DMT-2'Fluoro Phosphoramidites
5'-NPPOC Phosphoramidites
 Fast Deprotection Chemistries:
DNA
RNA
 Supports:
Controlled Pore Glass (CPG)
Columns
 Solvents:
Liquid Reagents
Activator 42™
 Modifications:
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
6-FAM
 Linkers:
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
RNA Phosphoramidites with Fast Deprotection Chemistry  
  Download Literature:
  •  User Instructions
Fast Deprotecton RNA Phosphoramidites
(770k pdf)
  •  Standard RNA Phosphoramidites
  View Structure:
  •  rA(tac) Phosphoramidite
  •  rC(tac) Phosphoramidite
  •  rG(tac) Phosphoramidite
  •  rU Phosphoramidite
SAFC Proligo Reagents offers RNA phosphoramidite monomers with Fast Deprotection Chemistry (tert-butylphenoxyacetyl (TAC) group).
Advantages of synthesis with TAC-protected RNA phosphoramidites
Increased synthesis yield and reduced purification efforts
Dramatically reduced deprotection times
Minimized exposure to alkaline deprotection medium
Minimized chain degradation during deprotection
RNA Phosphoramidites
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
A211081-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 x 0.5g
C213081-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 x 0.5g
G211081-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 x 0.5g
U211081-01 DMT-2′-O-TBDMS-rU amidite 1 x 0.5g
Compatible with ABI™ Instruments
Product No. Description Unit
A212031-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 x 0.5g
A212061-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 x 1 g
C212031-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 x 0.5g
C212061-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 x 1 g
G212031-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 x 0.5g
G212061-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 x 1 g
U211031-01 DMT-2′-O-TBDMS-rU amidite 1 x 0.5g
U211061-01 DMT-2′-O-TBDMS-rU amidite 1 x 1 g
  Other quantities and packaging are available upon request.
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP.
©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.
To order or speak with a Customer Service Representative, contact your local SAFC office.