Extranet 
Welcome to SAFC Supply Solutions
ssH Linker
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites:
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
2'Fluoro Phosphoramidites
DMT-2'Fluoro Phosphoramidites
5'-NPPOC Phosphoramidites
 Fast Deprotection Chemistries:
DNA
RNA
 Supports:
Controlled Pore Glass (CPG)
Columns
 Solvents:
Liquid Reagents
Activator 42™
 Modifications:
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
6-FAM
 Linkers:
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
Phosphoramidite technology is ideally suited for the modification of synthetic oligonucleotides with covalently attached linkers. SAFC Proligo Reagents™ provides ready to use linkers for DNA/RNA synthesis machines as ß-cyanoethyl phosphoramidites.
Amino linkers are particularly suitable for the conjugation of biotin, fluorescent dyes or other modifiers and reporter groups to the 5' end of oligonucleotides, or for the attachment of oligonucleotides to microarrays.
In addition to established linkers MMT*-protected amino linker : ssH-Linker
  Download Literature:
  •  User Instructions
ssH-Linker
(199k pdf)
  •  Product Information
ssH-Linker
(201k pdf)
  View Structure:
  •  ssH-Linker
ssH-Linker
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Quantity
M010982-01 ssH-Linker 1 x 0.25g
Compatible with ABI® Instruments
Product No. Description Unit
M010932-01 ssH-Linker 1 x 0.25g
Advantages over conventional amino linkers:
  • Deprotection of the amino-group under exceptionally mild conditions which avoid depurination side reaction
  • Very high labelling efficiency
  • Better trityl-on purification efficiency
General features:
  • Excellent coupling efficiency
  • High solubility in acetonitrile
  • Coupling through standard synthesis protocols, identical to the coupling of DNA monomer phosphoramidites
  • Used with standard deblock-, activator-, oxidizer- and capping-solutions
  • Features a lipophilic MMT-group to support the purification of the oligonucleotide synthesis product with reversed-phase cartridges or by HPLC
  • Permits measurement of the coupling efficiency through colorimetric MMT-assays
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP.
©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.
* monomethoxytrityl
ssH-Linker is covered by the patents / patent applications JP 2005-217026 and PCT/JP2005/021135.
To order or speak with a Customer Service Representative, contact your local SAFC office.