Extranet 
Welcome to SAFC Supply Solutions
Standard RNA Phosphoramidite
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites:
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
2'Fluoro Phosphoramidites
DMT-2'Fluoro Phosphoramidites
5'-NPPOC Phosphoramidites
 Fast Deprotection Chemistries:
DNA
RNA
 Supports:
Controlled Pore Glass (CPG)
Columns
 Solvents:
Liquid Reagents
Activator 42™
 Modifications:
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
6-FAM
 Linkers:
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
RNA plays a pivotal role in biological systems due to its numerous function in the transfer and processing of genetic information. The unique properties of RNA have stimulated the development of a variety of applications in diagnostics and therapeutics, as well as in basic molecular biology research where RNA can be used as:
Catalytic agents (ribozymes)
Affinity ligands (aptamers)
Agents to induce gene silencing (RNA interference)
Today, RNA interference (RNAi) has become a popular tool for the sequence-specific inhibition of gene expression and can be used in target validation and other drug development techniques. The most convenient method to provide sequence specific RNA oligonucleotides is chemical synthesis on a solid support with RNA phosphoramidites and RNA CPG, analogous to DNA synthesis.
Key features of Standard RNA Amidites
Industry standard 2'-TBDMS protective group
Lot-to-lot consistent purity and performance
Compatible with deprotection methods based on
methylamine or AMA
Standard RNA Amidites provide excellent coupling
results when used together with ETT or BTT as activator;
best results are obtained with Activator 42™1
Capping with standard acetic anhydride capping
reagent rather than with Fast Deprotection Cap A
Manufactured under a certified ISO 9001 quality system
  Download Literature:
  •  User Instructions
Standard RNA Phosphoramidites
(67k pdf)
  •  RNA Phosphoramidites
  View Structure:
  •  rA(bz) Amidite
  •  rC(ac) Amidite
  •  rG(ib) Amidite
  •  rU Amidite
RNA Phosphoramidites
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
A212081-01 DMT-2'O-TBDMS-rA(bz) Amidite 1 x 0.5g
C212081-01 DMT-2'O-TBDMS-rC(ac) Amidite 1 x 0.5g
G212081-01 DMT-2'O-TBDMS-rG(ib) Amidite 1 x 0.5g
U211081-01 DMT-2'O-TBDMS-rU Amidite 1 x 0.5g
Compatible with ABI™ Instruments
Product No. Description Unit
A212031-01 DMT-2'O-TBDMS-rA(bz) Amidite 1 x 0.5g
A212061-01 DMT-2'O-TBDMS-rA(bz) Amidite 1 x 1 g
C212031-01 DMT-2'O-TBDMS-rC(ac) Amidite 1 x 0.5g
C212061-01 DMT-2'O-TBDMS-rC(ac) Amidite 1 x 1 g
G212031-01 DMT-2'O-TBDMS-rG(ib) Amidite 1 x 0.5g
G212061-01 DMT-2'O-TBDMS-rG(ib) Amidite 1 x 1 g
U211031-01 DMT-2'O-TBDMS-rU Amidite 1 x 0.5g
U211061-01 DMT-2'O-TBDMS-rU Amidite 1 x 1 g
  Other quantities and packaging are available upon request.
1Activator 42 = 5-(3,5-Bis(trifluoromethyl)phenyl)-1H-tetrazole, a very powerful activator for RNA- and DNA phosphoramidites, commercially available through SAFC Proligo Reagents.
To order or speak with a Customer Service Representative, contact your local SAFC office.
SAFC and SAFC Proligo Reagents are registered trademarks of Sigma-Aldrich Biotechnology, LP.
©2009 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.