Suzuki Coupling Reagents
SAFC, a leader in boron reagents and compounds, is positioned to be a competitive supplier of arylboronic acids and Suzuki coupling catalysts to meet increasing development and large scale demands. We are also able to manufacture a wide range of other boron compounds, including a new generation of borane reagents that are more stable, more eco-friendly, and more reactive for synthetic applications.
Suzuki Palladium-Catalyzed Cross-Coupling with Aryl Halides
Arylboronic acids, including heterocyclic boronic acids, are key reagents in Suzuki palladium-catalyzed cross-coupling with aryl halides.
The use of air- and water-stable arylboronic acids in Suzuki reactions is superior in many aspects to other routes, including organometallic and Stille coupling reactions. Besides increased stability, arylboronic acids offer low toxicity and limited side reactions as compared to alternate chemistries.
These attributes are particularly beneficial in making pharmaceutical intermediates and APIs.
Contact SAFC
Contract Manufacturing Inquiries
For product/service inquiries, quotes or information requests
Manufacturing Facilities