|
Website Help System
Oligo Upload
|
• Instructions to Order Only One Type of Oligo • Instructions to Order More Than One Type of Oligo • Plate Formats • Ordering/Modification Codes
Instructions to Order Only One Type of Oligo
- Select the product type and file format from the drop down menu
- Format Your Sequences
- MS Excel Format (Tab Delimited): Column A = Oligo Name, Column B = Sequence, Column C = Scale (or Fixed OD), Column D = Purification, Column E = Special Instructions (Optional)
Example:
| My Oligo1 |
[A488]ACTGTGCATATTTTCCATAGCGT |
0.2 |
HPLC |
| My Oligo2 |
ACTGTGCATACCCATAGCGATTTTTT |
0.2 |
HPLC |
| My Oligo3 |
ACTGTGCATACCATAGCGTATATATAT |
0.2 |
HPLC |
- csv file Format (Comma Separated): Oligo Name, Sequence, Scale, Purification, Special Instructions (Optional)
Example: My Oligo1,[A488]ACTGTGCATATTTTCCATAGCGT,0.2,HPLC My Oligo2,ACTGTGCATACCCATAGCGATTTTTT,0.2,HPLC My Oligo3,ACTGTGCATACCATAGCGTATATATAT,0.2,HPLC
- Define Product Packaging and Format. To find details about plate formats, click here.
- Items can be added to your shopping cart by clicking Add to Cart.
Back to Top
Instructions to Order More Than One Type of Oligo
- Select "Defined in Data—Supports multiple types in one upload" from the drop down menu
- Select file format from the drop down menu
- Format Your Sequences
- MS Excel Format (Tab Delimited): Column A = Oligo Type Code, Column B = Oligo Name, Column C = Sequence, Column D = Scale (or Fixed OD), Column E = Purification, Column F = Special Instructions (Optional)
Example:
| STD |
My Oligo1 |
[A488]ACTGTGCATATTTTCCATAGCGT |
0.2 |
HPLC |
| STD |
My Oligo2 |
ACTGTGCATACCCATAGCGATTTTTT |
0.2 |
HPLC |
| STD |
My Oligo3 |
ACTGTGCATACCATAGCGTATATATAT |
0.2 |
HPLC |
| BBP |
BigBand Primer 1 |
ACTGTGCATACCATAGCGTATATATAT |
0.05 |
DST |
- csv file Format (Comma Separated): Oligo Type Code, Oligo Name, Sequence, Scale, Purification, Special Instructions (Optional)
Example: STD,My Oligo1,[A488]ACTGTGCATATTTTCCATAGCGT,0.2,HPLC STD,My Oligo2,ACTGTGCATACCCATAGCGATTTTTT,0.2,HPLC STD,My Oligo3,ACTGTGCATACCATAGCGTATATATAT,0.2,HPLC BBP,My BigBand,ACTGTGCATACCATAGCGTATATATAT,0.05,DST
- Define Product Packaging and Format. To find details about plate formats, click here.
- Items can be added to your shopping cart by clicking Add to Cart.
Back to Top
Plate Formats
- MS Excel Format: Column A = Oligo Name, Column B = Sequence, Column C = Scale (or Fixed OD), Column D = Purification, Column E = Plate Name (Optional)
- csv file Format (Comma Separated): Oligo Name, Sequence, Scale, Purification, Plate Name (Optional)
96-Well Plate(s), 8-Channel
Back to Top
96-Well Plate(s), 12-Channel
Back to Top
Ordering Codes
| Product Type Description |
Oligo Type Code |
| DNA Oligos - modified and non-modified |
STD |
| Dual labeled fluorogenic probe |
DLFP |
| Molecular Beacon |
MB |
| BigBand™ Primer |
BBP |
| F@ST |
F@STSTOLIGO |
| E@SY |
E@SYOLIGO |
| WellRED |
WELLRED |
| Pure and Simple Primer |
PSP |
Back to Top
| Scale of Synthesis Description |
Scale Code |
| 0.025 µmol |
0.025 |
| 0.05 µmol |
0.05 |
| 0.2 µmol |
0.2 |
| 1 µmol |
1 |
| 10 µmol |
10 |
| 15 µmol |
15 |
Back to Top
| Fixed OD Description (Labeled Probes & Beacons) |
Fixed OD Code |
| 1 OD |
1 |
| 3 OD |
3 |
| 5 OD |
5 |
| 10 OD |
10 |
| 50 OD |
50 |
| 100 OD |
100 |
Back to Top
| Purification Description |
Purification Code |
| Deprotected and Desalted |
DST |
| Reverse Phase Cartridge Purification |
RP1 |
| HPLC |
HPLC |
| PAGE |
PAGE |
Back to Top
| 5' Modification Description |
5' Mod Code |
| Alexa A488 |
[A488] |
| Alexa A546 |
[A546] |
| 5'Amine C12 |
[AmC12] |
| Amine C6 TFA (Desalt, PAGE) |
[AmC6F] |
| 5' Amino C6 MMT (RP1, HPLC) |
[AmC6T] |
| Biotin |
[Btn] |
| Biotin TEG |
[BtnTg] |
| 5-Bromo dC |
[5BrdC] |
| 5-Bromo dU |
[5BrdU] |
| 8-Bromo dA |
[8BrdA] |
| 8-Bromo dG |
[8BrdG] |
| Carboxy-Rhodamine 6G |
[CRh6G] |
| CY 3 |
[Cy3] |
| CY 5 |
[Cy5] |
| CY 5.5 |
[Cy55] |
| deoxyInosine |
I |
| deoxyUridine |
U |
| Digoxigenin |
[DIG] |
| 6-FAM |
[6FAM] |
| Fluorescein |
[Flc] |
| HEX |
[HEX] |
| JOE |
[JOE] |
| 5' Phosphorylation |
[Phos] |
| ROX |
[ROX] |
| TAMRA |
[TAM] |
| TET |
[TET] |
| Texas Red |
[TxRd] |
| Thiol C6 S-S |
[ThiC6] |
| WellRED D2-PA |
[D2-PA] |
| WellRED D3-PA |
[D3-PA] |
| WellRED D4-PA |
[D4-PA] |
| SPECIAL (Please define) |
[SPECIAL] |
Back to Top
| Base/IUB Redundancy Description |
Base/IUB Code |
| A (adenine) |
A |
| C (cytosine) |
C |
| G (guanine) |
G |
| T (thymine) |
T |
| Phosphorothioate linkages |
* |
| 2'O-Methyl RNA |
[mA], [mG], [mC], [mU] |
| LNA Bases |
[+A], [+G], [+C], [+T] |
| DNA bases within RNA oligos |
[dA], [dC], [dG], [dT] |
| B = G+T+C |
B |
| D = G+A+T |
D |
| H = A+T+C |
H |
| K = G+T |
K |
| M = A+C |
M |
| N = A+C+G+T |
N |
| R = A+G |
R |
| S = G+C |
S |
| V = A+C+G |
V |
| W = A+T |
W |
| Y = C+T |
Y |
Back to Top
| Internal Modification Description |
Internal Mod Code |
| Amino C2 dT |
[AmC2T] |
| Amino C6 dT |
[AmC6T] |
| Biotin TEG |
[BtnTg] |
| Biotin dT |
[BtndT] |
| 5-Bromo dC |
[5BrdC] |
| 5-Bromo dU |
[5BrdU] |
| 8-Bromo dA |
[8BrdA] |
| 8-Bromo dG |
[8BrdG] |
| deoxylnosine |
I |
| deoyxUridine |
U |
| Fluorescein dT |
[FlcdT] |
| 5 Methyl dC |
[5MedC] |
| SPECIAL (Please define) |
[SPECIAL] |
Back to Top
| 3' Modification Description |
3' Mod Code |
| Alexa A488 |
[A488] |
| Alexa A546 |
[A546] |
| 3'Amine C3 |
[AmC3] |
| 3'Amine C7 |
[AmC7] |
| Biotin TEG |
[BtnTg] |
| CY3 |
[Cy3] |
| CY5 |
[Cy5] |
| deoxylnosine |
I |
| deoxyUridine |
U |
| Digoxigenin |
[DIG] |
| Fluorescein |
[Flc] |
| JOE |
[JOE] |
| 3' Phosphorylation |
[Phos] |
| ROX |
[ROX] |
| Spacer C3 |
[SpcC3] |
| TAMRA |
[TAM] |
| Texas Red |
[TxRd] |
| Thiol C3 |
[ThiC3] |
| SPECIAL (Please define) |
[SPECIAL] |