Sigma-Aldrich
Welcome > Risk and Safety Statements
Website Help System
Risk and Safety Statements
European Union Commission Directive 2001/59/EC defines hazard codes and phrases
for classification, packaging and labeling of dangerous substances.
For complete hazard and safety information always refer to the MSDS.
RISK AND SAFETY STATEMENTS
For your convenience Sigma's labels include Risk and Safety numbers.
Please refer to the following table for the respective Risk and Safety Statements
for these numbers and combinations of numbers. In addition, Fluka Brand products contain F codes to assist with Risk and Safety information. Fluka F codes are also outlined below.
Risk Phrases
Indication of Particular Risks
Combination of Particular Risks
Safety Precautions
Indication of Safety Precautions
Combination of Safety Precautions
Fluka Brand F Codes
  ______________________________________________________________________
Pictograms and Hazard Codes
Risk Phrases
Indication of Particular Risks
R1: Explosive when dry
2: Risk of explosion by shock, friction, fire or other sources of ignition
3: Extreme risk of explosion by shock, friction, fire or other sources of ignition
4: Forms very sensitive explosive metallic compounds
5: Heating may cause an explosion
6: Explosive with or without contact with air
7: May cause fire
8: Contact with combustible material may cause fire
9: Explosive when mixed with combustible material
10: Flammable
11: Highly Flammable
12: Extremely Flammable
14: Reacts violently with water
15: Contact with water liberates extremely flammable gases
16: Explosive when mixed with oxidizing substances
17: Spontaneously flammable in air
18: In use may form flammable/explosive vapor-air mixture
19: May form explosive peroxides
20: Harmful by inhalation
21: Harmful in contact with skin
22: Harmful if swallowed
23: Toxic by inhalation
24: Toxic in contact with skin
25: Toxic if swallowed
26: Very Toxic by inhalation
27: Very Toxic in contact with skin
28: Very Toxic if swallowed
29: Contact with water liberates toxic gas
30: Can become highly flammable in use
31: Contact with acids liberates toxic gas
32: Contact with acids liberates very toxic gas
33: Danger of cumulative effects
34: Causes burns
35: Causes severe burns
36: Irritating to the eyes
37: Irritating to the respiratory system
38: Irritating to the skin
39: Danger of very serious irreversible effects
40: Limited evidence of a carcinogenic effect
41: Risk of serious damage to eyes
42: May cause sensitization by inhalation
43: May cause sensitization by skin contact
44: Risk of explosion if heated under confinement
45: May cause cancer
46: May cause heritable genetic damage
48: Danger of serious damage to health by prolonged exposure
49: May cause cancer by inhalation
50: Very Toxic to aquatic organisms
51: Toxic to aquatic organisms
52: Harmful to aquatic organisms
53: May cause long-term adverse effects in the aquatic environment
54: Toxic to flora
55: Toxic to fauna
56: Toxic to soil organisms
57: Toxic to bees
58: May cause long-term adverse effects in the environment
59: Dangerous for the ozone layer
60: May impair fertility
61: May cause harm to the unborn child
62: Possible risk of impaired fertility
63: Possible risk of harm to the unborn child
64: May cause harm to breast-fed babies
65: Harmful: May cause lung damage if swallowed
66: Repeated exposure may cause skin dryness or cracking
67: Vapors may cause drowsiness and dizziness
68: Possible risk of irreversible effects
Risk Phrases
Combination of Particular Risks
14/15: Reacts violently with water, liberating extremely flammable gases
15/29: Contact with water liberates toxic, extremely flammable gas
20/21: Harmful by inhalation and in contact with skin
20/21/22: Harmful by inhalation, in contact with skin and if swallowed
20/22: Harmful by inhalation and if swallowed
21/22: Harmful in contact with skin and if swallowed
23/24: Toxic by inhalation and in contact with skin
23/24/25: Toxic by inhalation, in contact with skin and if swallowed
23/25: Toxic by inhalation and if swallowed
24/25: Toxic in contact with skin and if swallowed
26/27: Very Toxic by inhalation and in contact with skin
26/27/28: Very Toxic by inhalation, in contact with skin and if swallowed
26/28: Very Toxic by inhalation and if swallowed
27/28: Very Toxic in contact with skin and if swallowed
36/37: Irritating to eyes and respiratory system
36/37/38: Irritating to eyes, respiratory system and skin
36/38: Irritating to eyes and skin
37/38: Irritating to respiratory system and skin
39/23: Toxic: danger of very serious irreversible effects through inhalation
39/23/24: Toxic: danger of very serious irreversible effects through inhalation in contact with skin
39/23/24/25: Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if swallowed
39/23/25: Toxic: danger of very serious irreversible effects through inhalation and if swallowed
39/24: Toxic: danger of very serious irreversible effects in contact with skin
39/24/25: Toxic: danger of very serious irreversible effects in contact with skin and if swallowed
39/25: Toxic: danger of very serious irreversible effects if swallowed
39/26: Very Toxic: danger of very serious irreversible effects through inhalation
39/26/27: Very Toxic: danger of very serious irreversible effects through inhalation and in contact with skin
39/26/27/28: Very Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if swallowed
39/26/28: Very Toxic: danger of very serious irreversible effects through inhalation and if swallowed
39/27: Very Toxic: danger of very serious irreversible effects in contact with skin
39/27/28: Very Toxic: danger of very serious irreversible effects in contact with skin and if swallowed
39/28: Very Toxic: danger of very serious irreversible effects if swallowed
42/43: May cause sensitization by inhalation and skin contact
48/20: Harmful: danger of serious damage to health by prolonged exposure through inhalation
48/20/21: Harmful: danger of serious damage to health by prolonged exposure through inhalation and in contact with skin
48/20/21/22: Harmful: danger of serious damage to health by prolonged exposure through inhalation, and in contact with skin and if swallowed
48/20/22: Harmful: danger of serious damage to health by prolonged exposure through inhalation and if swallowed
48/21: Harmful: danger of serious damage to health by prolonged exposure in contact with skin
48/21/22: Harmful: danger of serious damage to health by prolonged exposure in contact with skin and if swallowed
48/22: Harmful: danger of serious damage to health by prolonged exposure if swallowed
48/23: Toxic: danger of serious damage to health by prolonged exposure through inhalation
48/23/24: Toxic: danger of serious damage to health by prolonged exposure through inhalation and in contact with skin
48/23/24/25: Toxic: danger of serious damage to health by prolonged exposure through inhalation, in contact with skin and if swallowed
48/23/25: Toxic: danger of serious damage to health by prolonged exposure through inhalation and if swallowed
48/24: Toxic: danger of serious damage to health by prolonged exposure in contact with skin
48/24/25: Toxic: danger of serious damage to health by prolonged exposure in contact with skin and if swallowed
48/25: Toxic: danger of serious damage to health by prolonged exposure if swallowed
50/53: Very Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment
51/53: Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment
52/53: Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment
68/20: Harmful: possible risk of irreversible effects through inhalation
68/20/21: Harmful: possible risk of irreversible effects through inhalation and in contact with skin
68/20/21/22: Harmful: possible risk of irreversible effects through inhalation, in contact with skin and if swallowed
68/20/22: Harmful: possible risk of irreversible effects through inhalation and if swallowed
68/22: Harmful: possible risk of irreversible effects if swallowed
68/21: Harmful: possible risk of irreversible effects in contact with skin
68/21/22: Harmful: possible risk of irreversible effects in contact with skin and if swallowed
68/22: Harmful: possible risk of irreversible effects if swallowed
Back to Top
Safety Phrases
Indication of Safety Precautions
S1: Keep locked up
2: Keep out of the reach of children
3: Keep in a cool place
4: Keep away from living quarters
5: Keep contents under ... (appropriate liquid to be specified by the manufacturer)
6: Keep under ... (inert gas to be specified by the manufacturer)
7: Keep container tightly closed
8: Keep container dry
9: Keep container in a well-ventilated place
12: Do not keep the container sealed
13: Keep away from food, drink and animal feeding stuffs
14: Keep away from ... (incompatible materials to be indicated by the manufacturer)
15: Keep away from heat
16: Keep away from sources of ignition - No smoking
17: Keep away from combustible material
18: Handle and open container with care
20: When using, do not eat or drink
21: When using, do not smoke
22: Do not breathe dust
23: Do not breathe gas/fumes/vapor/spray (appropriate wording to be specified by the manufacturer)
24: Avoid contact with skin
25: Avoid contact with eyes
26: In case of contact with eyes, rinse immediately with plenty of water and seek medical advice
27: Take off immediately all contaminated clothing
28: After contact with skin, wash immediately with plenty of ... (to be specified by the manufacturer)
29: Do not empty into drains
30: Never add water to this product
33: Take precautionary measures against static discharges
35: This material and its container must be disposed of in a safe way
36: Wear suitable protective clothing
37: Wear suitable gloves
38: In case of insufficient ventilation, wear suitable respiratory equipment
39: Wear eye/face protection
40: To clean the floor and all objects contaminated by this material use ... (to be specified by the manufacturer)
41: In case of fire and/or explosion do not breathe fumes
42: During fumigation/spraying wear suitable respiratory equipment (appropriate wording to be specified)
43: In case of fire, use ... (indicate in the space the precise type of fire-fighting equipment. If water increases the risk add - Never use water)
45: In case of accident or if you feel unwell, seek medical advice immediately (show label where possible)
46: If swallowed, seek medical advice immediately and show this container or label
47: Keep at temperature not exceeding ... E C (to be specified by the manufacturer)
48: Keep wetted with ... (appropriate material to be specified by the manufacturer)
49: Keep only in the original container
50: Do not mix with ... (to be specified by the manufacturer)
51: Use only in well-ventilated areas
52: Not recommended for interior use on large surface areas
53: Avoid exposure - obtain special instruction before use
56: Dispose of this material and its container to hazardous or special waste collection point
57: Use appropriate container to avoid environmental contamination
59: Refer to manufacturer/supplier for information on recovery/recycling
60: This material and/or its container must be disposed of as hazardous waste
61: Avoid release to the environment. Refer to special instructions safety data sheet
62: If swallowed, do not induce vomiting: seek medical advice immediately and show this container or label
63: In case of accident by inhalation, remove casualty to fresh air and keep at rest
64: If swallowed, rinse mouth with water (only if the person is conscious)
Back to Top
Safety Phrases
Combination of Safety Precautions
1/2: Keep locked up and out of the reach of children
3/7: Keep container tightly closed in a cool place
3/9/14: Keep in a cool well-ventilated place away from ... (incompatible materials to be indicated by manufacturer)
3/9/14/49: Keep only in the original container in a cool well-ventilated place away from ... (incompatible materials to be indicated by manufacturer)
3/9/49: Keep only in the original container in a cool well-ventilated place
3/14: Keep in a cool place away from ... (incompatible materials to be indicated by the manufacturer)
7/8: Keep container tightly closed and dry
7/9: Keep container tightly closed and in a well-ventilated place
7/47: Keep container tightly closed and at a temperature not exceeding ... °C (to be specified by manufacturer)
20/21: When using, do not eat, drink or smoke
24/25: Avoid contact with skin and eyes
27/28: After contact with skin, take off immediately all contaminated clothing and wash imediately with plenty of.... (to be specified by the manufacturer)
29/35: Do not empty into drains, dispose of this container and it's material in a safe way
29/56: Do not empty into drains, dispose of this material and its container to hazardous or special waste-collection point
36/37: Wear suitable protective clothing and gloves
36/37/39: Wear suitable protective clothing, gloves and eye/face protection
36/39: Wear suitable protective clothing and eye/face protection
37/39: Wear suitable gloves and eye/face protection
47/49: Keep only in the original container at temperature not exceeding ... °C (to be specified by manufacturer)
Back to Top
FLUKA BRAND F CODES
  F 1 Sensitive to air and humidity.
F 2 Sensitive to air and CO2.
F 3 Hygroscopic.
F 4 Do not heat over …°C.
  1 = 10 °C
  2 = 20 °C
  3 = 30 °C
  4 = 40 °C
  5 = 50 °C
  6 = 60 °C
  7 = 70 °C
  8 = 80 °C
  9 = 90 °C
  10 = 100 °C
  11 = 110 °C
  12 = 120 °C
  13 = 130 °C
  14 = 140 °C
  15 = 150 °C
F 5 Dry material may explode.
F 6 Reacts with acids.
F 7 Keep under CO2.
F 8 Photosensitive.
F 9 Keep under nitrogen.
F 10 Keep under argon.
F 11 Keep under inert gas.
F 12 Shake before use.
F 13 Malodorous.
F 14 Carefully observe warning label.
F 15 Limited shelf life.
F 16 Decomposes easily.
F 17 May decompose by explosion.
F 18 Must be handled by instructed people only.
F 19 Lachrymator.
F 20 Not to be swallowed.
F 21 Sensitive to humidity.
F 22 Keep dust-free in the dark.
F 23 Sensitive to air.
F 24 Spontaneously flammable with alcohol.
F 25 Keep under CO.
F 26 Before distillation test for absence of peroxides.
F 27 Cool before opening.
F 28 May become cloudy (Polyformaldehyde).
F 29 Spontaneously flammable when dry.
F 30 Sediment unavoidable.
F 31 Do not puncture.
F 32 Avoid contact with heavy metal salts
        especially iron salts.
F 33 Causes fall in blood pressure.
F 34 Sensitive to CO2.
F 35 Narcotic effects.
F 36 Extremely sensitive to air.
Back to Top
PICTOGRAMS AND HAZARD CODES
  biohazard B Biohazard flammable
F  Highly Flammable
F+  Extremely Flammable
  oxidizing1 O Oxidizing
corrosive C Corrosive harmful
Xn  Harmful
Xi  Irritant
radiation R Radioactive
explosive E Explosive envirodanger N
 Dangerous for the
 environment
toxic
T  Toxic
T+  Very Toxic
Back to Top
Sigma Aldrich Logo
 © 2009 Sigma-Aldrich Co. All Rights Reserved.  Reproduction of any materials from the site is strictly forbidden without permission.  Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.