Issue 1 Food Chemistry
Featured Applications
Analytical testing is used in the production of worldwide food supply. This testing primarily serves two purposes: to determine nutritional value and quality, and to guarantee safety. Sigma-Aldrich®, through its Supelco® brand, is very active in product development for food and beverage analysis.
Food Chemistry
Many food companies are currently and may soon to be under more pressure from the Food and Drug Administration to make their foods healthier. Since 2006, the FDA indicates requirements to include trans-fat content on nutritional labels. However, they are often, but not always, found in products.(source)
In the following application, our scientists developed a procedure to determine trans-fat levels in foods. This procedure involves extraction, derivatization, fractionation and GC analysis.
Comprehensive Determination of Trans Fats in Cookies
Using Silver-Ion SPE Fractionation, SP™-2560 and SLB®-IL111 GC Columns …more
1. Silverion (Ag-ion) fractionation is applied to the FAMEs to separate the saturates and C18 trans monoenes in fraction 1, the cis monoenes in fraction 2, and the dienes in fraction 3.
2. GC analysis of each fraction is then performed on an ionic liquid GC column. This extremely polar GC capillary column was the world’s first commercial column to rate over 100 on our GC column polarity scale. As such, it has the most orthogonal selectivity compared to commonly used non-polar and intermediate polar columns, providing increased selectivity for polar and polarizable analytes. The 100 m version is a great complementary column to the SP-2560. …more
Analytical Solutions: Gas Chromatography
Ionic Liquid Columns for Gas Chromatography
What exactly are Ionic Liquids? How do they perform and improve Gas Chromatography? Get your answers here!
Ionic Liquid Capillary Columns for the Analysis of FAME Isomers
Analyses of fatty acid methyl esters (FAMEs) are continuing to gain importance as more research is focusing on their biomedical impacts. Our two new phases are SLB®-IL60 with a PEG like selectivity and the SLB-IL111 with highly polar selectivity. We will compare and contrast the selectivity of the ionic liquid phases with polymeric based phases for various FAME samples. …read more on SLB®-IL60 and SLB-IL111
USP Methods and Sigma-Aldrich® GC Columns
U.S. Pharmacopeial Convention (USP) methods typically cover pharmaceutical products, which may be necessary in F&B testing. Here are the USP codes and our recommended Supelco capillary GC columns
Click here
Food Chemistry Analytical Workflow for F&B & F&F Industry
Sigma Aldrich is actively expanding our product offerings to our Food & Beverage and Flavours & Fragrances customers!
Food Chemistry Product Portfolio
Please view our F&B Food Chemistry product portfolio and click on the respective product links for more information!
Carbohydrates &
Dietary Fiber
Enzymatic Kits
Dietary Fiber Kits
ApHera HPLC Columns
SUPELCOGEL HPLC Columns
Ascentis Express  HPLC Columns (HILIC, OH5, F5)
Analytical Standards & CRMs
HPLC Analytical Reagents & Solvents
Fatty Acids,
FAMEs,
Sterols &
Edible Oils
Derivatization Reagents
LLE Extraction Glassware
Discovery® Ag-Ion SPE Tubes
Nukol Capillary GC Columns
Omegawax® & Ionic Liquids (SLB-IL59, 60, 111) Capillary GC Columns
Equity-1 Capillary GC Columns
MET-Biodiesel Capillary GC Columns
SP-2560 & SP-2380 Capillary GC Columns
SAC-5 Capillary  GC Columns
Ascentis® Express HPLC Columns
Analytical Standards & CRMs
- Characterized Ref. Oils
GC Solvents
HPLC Analytical Reagents & Solvents
IR & NMR Spectroscopy
Proteins
Extraction Glassware
Derivatization Reagents
Trypsin Digestion Reagent
SUPELCOSIL LC-DABS HPLC Columns
Astec CHIROBIOTIC TAG HPLC Columns
Astec CHIROBIOTIC T HPLC Columns
Astec CHIROBIOTIC T2 HPLC Columns
Discovery C8 & C18 HPLC Columns
HPLC Columns for Biopolymers
Analytical Standards & CRMs
HPLC Analytical Reagents & Solvents
Equipment for Kjeldahl Method
Kjeldahl Catalysts & Oxidants for Digestion
Reference Flavours and Oil Standards
Flavors & Fragrances
We are also constantly increasing our selection of reference flavours and oils standards. Check them out here.
Solutions from Formulation to Testing
Flavors & Fragrances Guide Try Vitroids Mycotoxin Analysis Brochure
Download our Application Guide featuring a selection of new products.
Learn how you can save time and money by using Vitroids. Download our Mycotoxin Analysis brochure for products and solutions along the mycotoxin analysis workflow.
Next issue: Coming up…
- Environmental Testing featuring Air monitoring and water testing
- Dioxin Analysis, Environmental Standards and Proficiency Testing of Laboratories
- Air Sampling Devices Solutions
- F&F: Testing and Cosmetics Testing Analytical Workflows
And more..
Upcoming Launch:
1. Titan™ UHPLC Columns
Promotion
Up to 50% discount on selected HPLC solvents
Promotion  Extended till 30th April 2014
Part Number
Description
List Price
Extended
promo price
Discount
34966-4L
MeOH LCMS Grade
SGD 62.00
SGD 37.20
40%
34967-4L
ACN LCMS Grade
SGD 140.00
SGD 70.00
50%
Terms and conditions:
1.        A minimum order of SGD $150 must be placed in one PO to be entitled for this promotion
2.        Please send your orders to sapl@sial.com or reply to this email to be entitled.
3.        Only valid for Singapore customers, excluding resellers, till 30th April 2014 or while stocks last.
For more information:
Phone: (65) 6779 1200 | Fax: (65) 6779 1822 | Email: sapl@sial.com