ASTM D5501: GC Analysis of Ethanol Content in E85 Bioethanol on Petrocol® DH 150
ASTM D5501: GC Analysis of Ethanol Content in E85 Bioethanol on Petrocol® DH 150
Conditions
column Petrocol DH 150, 150 m x 0.25 mm I.D., 1.0 µm (24155)
oven 60 °C (15 min), 30 °C/min to 250 °C (23 min)
carrier gas helium, 24 cm/sec @ 60 °C
sample E85 denatured fuel ethanol
injection 0.5 µL, 150:1 split
liner 4 mm I.D., single taper
inj. temp. 300 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Alcohols
Featured Industry Biofuels
Legal Information Petrocol is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes