ASTM D5501: GC Analysis of Ethanol on Petrocol® DH 150
ASTM D5501: GC Analysis of Ethanol on Petrocol® DH 150
Conditions
column Petrocol DH 150, 150 m x 0.25 mm I.D., 1.0 µm (24155)
oven 60 C (15 min), 30 C/min to 250 C (23 min)
carrier gas helium, 24 cm/sec @ 60 C
sample Denatured Fuel Ethanol Solution 4, from ASTM D5501 Denatured Fuel Ethanol Standards Kit (40361-U)
injection 0.5 µL, 150:1 split
liner 4 mm I.D., single taper
inj. temp. 300 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Alcohols
Featured Industry Biofuels
Legal Information Petrocol is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Ethanol
  • Methanol
  • n-Heptane