EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms
EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C (3 min), 25 C/min to 250 °C (3 min), 4 °C/min to 325 C (5 min)
carrier gas helium, 0.9 mL/min constant
sample 26 analytes at 50 ppb each in methylene chloride
injection 0.5 µL, pulsed (15 psi until 0.20 min) splitless (1.0 min)
liner 2 mm I.D. FocusLiner(TM) inlet liner with taper
inj. temp. 300 °C
MSD interface 300 °C / 200 °C (source/quads)
scan range SIM
Description
Categories Analytical Chromatography, GC Applications, PAHs, Pesticides, Hydrocarbons
Featured Industry Agriculture
Food and Beverages
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1'-(2,2,2-Trichloroethane-1,1-diyl)bis(4-chlorobenzene) (2,4'-DDT)
  • 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDT)
  • 5-Methylchrysene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[a]pyrene-d12
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[j]fluoranthene
  • Benzo[k]fluoranthene
  • Chrysene
  • Cyclopenta[c,d]pyrene
  • Dibenzo[a,e]pyrene
  • Dibenzo[a,h]anthracene
  • Dibenzo[a,h]pyrene
  • Dibenzo[a,i]pyrene
  • Dibenzo[a,l]pyrene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • m-Xylene
  • Naphthalene-d8
  • o-Xylene
  • Phenanthrene
  • p-Xylene
  • Styrene