GC Analysis / Mass Spectrum of Benzo[g,h,i]Perylene (elutes at 325 °C) on SLB®-5ms, from US EPA Method 8270
GC Analysis / Mass Spectrum of Benzo[g,h,i]Perylene (elutes at 325 °C) on SLB®-5ms, from US EPA Method 8270
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 40 °C (3 min), 20 °C/min to 100 °C, 10 °C/min to 200 °C, 30 °C/min to 325 °C (5 min)
carrier gas helium, pressure programmed, 20 psi (0 min), 99 psi/min to 80 psi (0 min), 99 psi/min to 16.5 psi (3 min), 99 psi/min to 25 psi (hold remainder of run)
sample 50 ng on-column of a 72 component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 40 ng on-column)
injection 1.0 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
MSD interface 325 °C
scan range 40-450 amu
Description
Categories Analytical Chromatography, GC Applications, Semivolatiles
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Benzo[g,h,i]perylene