GC Analysis of 1,2,3,4-Tetrahydro-1-Naphthylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-DA
GC Analysis of 1,2,3,4-Tetrahydro-1-Naphthylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-DA
Conditions
column Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 µm (72023AST)
oven 140 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: 1,2,3,4-tetrahydro-1-naphthylamine enantiomers (N-TFA derivatives)
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Anilines, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes